Skip to Content
Merck
All Photos(1)

Key Documents

SAB1300349

Sigma-Aldrich

Anti-Mouse Pcp2 (C-term) antibody produced in rabbit

saturated ammonium sulfate (SAS) precipitated, buffered aqueous solution

Synonym(s):

Anti-Pcp-2, Anti-Pcp2, Anti-Protein PCD-5, Anti-Purkinje cell protein 2, Anti-Purkinje cell-specific protein L7

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€374.00

€374.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μG
€374.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€374.00


Please contact Customer Service for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

saturated ammonium sulfate (SAS) precipitated

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

mouse

technique(s)

immunohistochemistry: 1:50-1:100
indirect ELISA: 1:1000

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

mouse ... Pcp2(18545)

General description

Pcp2 may function as a cell-type specific modulator for G protein-mediated cell signaling. PCP2 (Purkinje cell protein 2) codes for a protein belonging to the GoLoco domain-containing family. Pcp2 is specifically localized to cerebellum in the Purkinje cells and in retinal bipolar cells. From Pcp2 protein structural analysis, it is known that Pcp2 contains two GoLoco motifs.

Immunogen

PCP2 (NP_032816, )
This antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide selected from the C-terminal region of mouse Pcp2.

Biochem/physiol Actions

Pcp2 might serve as an inhibitor of guanine nucleotide dissociation.

Physical form

Purified polyclonal antibody supplied in PBS with 0.09% (W/V) sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Retinal ON bipolar cells express a new PCP2 splice variant that accelerates the light response.
Xu Y, et al.
The Journal of Neuroscience, 28(36), 8873-8884 (2008)
G-protein alpha subunit interaction and guanine nucleotide dissociation inhibitor activity of the dual GoLoco motif protein PCP-2 (Purkinje cell protein-2).
Willard F S, et al.
Cellular Signalling, 18(8), 1226-1234 (2006)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service