Skip to Content
Merck
All Photos(1)

Key Documents

SAB1300178

Sigma-Aldrich

Anti-Nematode SUMO (C-term) antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€318.00

€318.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μG
€318.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€318.00


Please contact Customer Service for Availability

biological source

rabbit

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect ELISA: 1:1000

NCBI accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... smo-1(266820)

General description

Covalent attachment of one protein to another is one of the more prominent posttranslational modifications in respects to size and ubiquity – to which eukaryotic proteins are subject. Ubiquitin is the most familiar of the protein modifiers and its activation and transfer to target proteins has been studied for over two decades. Recently a new group of ubiquitin-like (Ubl) proteins have come to light. One of the most intriguing of them is SUMO (small ubiquitin-like modifier, ~12kDa) also known as Sentrin. SUMO family has been described in vertebrates: SUMO-1 and the closest homologs SUMO-2 and SUMO-3. SUMO have been shown to bind and regulate mammalian SP-RINGs (such as Mdm2, PIAS and PML), RanGAP1, RanBP2, p53, p73, HIPK2, TEL, c-Jun, Fas, Daxx, TNFRI, Topo-I, Topo-II, WRN, Sp100, IkB-alpha, Androgen receptor (AR), GLUT1/4, Drosophila Ttk69, Dorsal, CaMK, yeast Septins, and viral CMV-IE1/2, EBV-BZLF1, HPV/BPV-E1. These bindings implicate SUMO in the stabilization of the target proteins and/or their localization to subcellular complexes. SUMO research enters now an exciting phase with a promise to help understanding how cells orchestrate the complexities of rapidly regulating protein level and activity. This antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide selected from the C-terminal region of nematode SUMO.

Immunogen

SUMO (NP_490842, 43-77)
This antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide selected from the C-terminal region of nematode SUMO.

Physical form

Purified polyclonal antibody supplied in PBS with 0.09% (W/V) sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service