S9262
Tin(II) chloride dihydrate
suitable for cell culture, ≥95%
Synonym(s):
Stannous chloride dihydrate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
SnCl2 · 2H2O
CAS Number:
Molecular Weight:
225.65
EC Number:
MDL number:
UNSPSC Code:
12352207
Recommended Products
Assay
≥95%
technique(s)
cell culture | mammalian: suitable
bp
652 °C (lit.)
mp
37-38 °C (dec.) (lit.)
SMILES string
O.O.Cl[SnH2]Cl
InChI
1S/2ClH.2H2O.Sn/h2*1H;2*1H2;/q;;;;+2/p-2
InChI key
FWPIDFUJEMBDLS-UHFFFAOYSA-L
Looking for similar products? Visit Product Comparison Guide
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service