Skip to Content
Merck
All Photos(1)

Key Documents

S8649

Sigma-Aldrich

Sar-Arg-Gly-Asp-Ser-Pro

≥97% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H39N9O10
CAS Number:
Molecular Weight:
601.61
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥97% (HPLC)

storage temp.

−20°C

SMILES string

CNCC(=O)NC(CCCNC(N)=N)C(=O)NCC(=O)NC(CC(O)=O)C(=O)NC(CO)C(=O)N1CCCC1C(O)=O

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P Wisniowski et al.
The Journal of laboratory and clinical medicine, 125(1), 38-45 (1995-01-01)
Pneumocystis carinii pneumonia is a major opportunistic infection in patients with the acquired immune deficiency syndrome. P. carinii attachment to alveolar epithelial cells is considered necessary for growth and replication of the organism. Recent studies have focused on the role

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service