Skip to Content
Merck
All Photos(1)

Documents

S7822

Sigma-Aldrich

Anti-SMC6 antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-SMC6L1, Anti-Structural maintenance of chromosome 6, Anti-Structural maintenance of chromosomes 6-like 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 126 kDa

species reactivity

human, rat (predicted), mouse (predicted)

technique(s)

western blot: 1:250-1:500 using whole cell extract of HEK293-T expressing SMC6

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SMC6(79677)
mouse ... Smc6(67241)
rat ... Smc6l1(313961)

General description

Structural maintenance of chromosomes protein 6 (SCM6) belongs to the structural maintenance of chromosomes family. There are at least six SMC family members that form three heterodimers in specific combinations. SCM6 gene is mapped to human chromosome 2p24.2. Structurally, SCM6 has DNA-binding domains at the N and C termini, a long coiled-coil domain, and a hinge region.

Specificity

Anti-SMC6 (also known as SMC6L1) specifically recognizes SMC6.

Immunogen

synthetic peptide corresponding to amino acids 766-779 of human SMC6, conjugated to KLH via an N-terminal cysteine residue. This sequence differs by one amino acid in mouse, and by two amino acids in rat.

Application

Anti-SMC6 antibody produced in rabbit may be used in immunoblotting.

Biochem/physiol Actions

Structural maintenance of chromosomes protein 6 (SCM6) along with SMC5 is mainly involved in the cellular response to DNA damage in yeast as well as humans. The yeast SMC5/6 complex contains several non-SMC elements (NSE), including methyl methanesulfonate-sensitivity protein 21 (MMS21) which sumoylates components of the complex. The SMC5/6 complex is recruited to nuclease-induced double strand breaks(DSBs). This event is crucial for human SMC1/3 cohesion complex recruitment to DSBs. SCM6 is known to interact with the papillomavirus E2 protein. Mitotic aberration and abnormal distribution of topoisomerase IIα are associated with depleted levels of Smc5 and Smc6.

Physical form

Solution in 0.01 M phos­phate buffered saline, pH 7.4, containing 15 mM sodium azide.

Storage and Stability

For continuous use, store at 2-8 °C for up to one month. For extended storage, freeze in working aliquots. Repeated freezing and thawing, or storage in “frost-free” freezers, is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilutions should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Patrick Ryan Potts et al.
Nature structural & molecular biology, 14(7), 581-590 (2007-06-26)
Most cancer cells activate telomerase to elongate telomeres and achieve unlimited replicative potential. Some cancer cells cannot activate telomerase and use telomere homologous recombination (HR) to elongate telomeres, a mechanism termed alternative lengthening of telomeres (ALT). A hallmark of ALT
Ming Yu et al.
Epigenetics, 10(9), 803-809 (2015-07-18)
Aberrant DNA methylation is a common epigenetic alteration found in colorectal adenomas and cancers and plays a role in cancer initiation and progression. Aberrantly methylated DNA loci can also be found infrequently present in normal colon tissue, where they seem
Chul-Kee Park et al.
Neuropathology : official journal of the Japanese Society of Neuropathology, 29(4), 443-449 (2009-01-28)
We analyzed the methylation status of the O6-methylguanine-DNA methyltransferase (MGMT) promoter using a methylation-specific polymerase chain reaction (MSP) in glioblastoma patients treated with 1-(4-amino-2-methyl-5-pyrimidinyl)methyl-3-(2-chloroethyl)-3-nitrosourea (ACNU) plus cisplatin followed by radiation therapy. Forty-eight patients with interpretable MSP results were included in
Dongjun Jeong et al.
Molecular medicine reports, 17(6), 8173-8179 (2018-04-26)
Loss of runt‑related transcription factor 3 (RUNX3) has been reported in various cancers, and one of the mechanisms mediating loss of RUNX3 expression is DNA methylation. However, the role of RUNX3 expression and its DNA methylation status as prognostic factors in
Jun Gao et al.
Oncology reports, 23(6), 1683-1692 (2010-04-30)
Pancreatic juice is a potential source of proteins associated with pancreatic cancer (PC) due to the proximity of ducts to tumor tissue. Therefore, screening of proteins in pancreatic juice from PC patients may identify new PC biomarkers. We analyzed pancreatic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service