Skip to Content
Merck
All Photos(1)

Key Documents

S7073

Sigma-Aldrich

SGK1 (60-end), active, GST tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
€420.00

€420.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 μG
€420.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

€420.00


Please contact Customer Service for Availability

Request a Bulk Order

recombinant

expressed in baculovirus infected Sf9 cells

Quality Level

product line

PRECISIO® Kinase

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

99-135 nmol/min·mg

mol wt

~73 kDa

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... SGK1(6446)

Biochem/physiol Actions

SGK1 is a member of the serum- and glucocorticoid-induced protein kinase that is activated in vitro by 3-phosphoinositide-dependent protein kinase-1 (PDK1) and in vivo in response to signals that activate phosphatidylinositol (PI) 3-kinase. SGK1 mRNA is expressed in all tissues and the level of SGK1 mRNA is increased by stimulation with serum or dexamethasone. SGK1 promotes cell survival by phosphorylating and inactivating FKHRL1. SGK and Akt display differences with respect to the efficacy with which they phosphorylate the three regulatory sites on FKHRL1.

Physical form

Supplied in 50 mM Tris-HCl, pH 7.5, with 150 mM NaCl, 0.2 5mM DTT, 0.1 mM EGTA, 0.1 mM EDTA, 0.1 mM PMSF, and 25% glycerol.

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Brunet et al.
Molecular and cellular biology, 21(3), 952-965 (2001-01-12)
Serum- and glucocorticoid-inducible kinases (SGKs) form a novel family of serine/threonine kinases that are activated in response to a variety of extracellular stimuli. SGKs are related to Akt (also called PKB), a serine/threonine kinase that plays a crucial role in
T Kobayashi et al.
The Biochemical journal, 344 Pt 1, 189-197 (1999-11-05)
The catalytic domain of serum- and glucocorticoid-induced protein kinase (SGK) is 54% identical with protein kinase B (PKB) and, like PKB, is activated in vitro by 3-phosphoinositide-dependent protein kinase-1 (PDK1) and in vivo in response to signals that activate phosphatidylinositol

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service