Skip to Content
Merck
All Photos(1)

Key Documents

S7031

Sigma-Aldrich

Storage box

polycarbonate, can be washed and autoclaved repeatedly

Synonym(s):

lab storage container, laboratory storage

Sign Into View Organizational & Contract Pricing

Select a Size

4 EA
€63.20

€63.20


Please contact Customer Service for Availability


Select a Size

Change View
4 EA
€63.20

About This Item

UNSPSC Code:
41123300
NACRES:
NB.22

€63.20


Please contact Customer Service for Availability

material

polycarbonate

packaging

pack of 4 ea

manufacturer/tradename

Nalgene® 5050-0001

L × W × H

5 1/4 in. × 5 1/4 in. × 2 in.

General description

Uncompartmented for storage of various size items in any environment from -196 to +121°C. Can be washed and autoclaved repeatedly.

Legal Information

Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service