Key Documents
S6565
Nunc® Lab-Tek® II - CC2 Chamber Slide system
2 wells, glass slide, 4.0 cm2/well, sterile
Synonym(s):
cell culture chamber, chamber slides, microscope slides, microscopy slides
Select a Size
1 PKG
€616.00
€616.00
Please contact Customer Service for Availability
Select a Size
Change View
1 PKG
€616.00
About This Item
UNSPSC Code:
41161502
NACRES:
NB.13
€616.00
Please contact Customer Service for Availability
material
polystyrene chamber
sterility
sterile
feature
CE marked
manufacturer/tradename
Nunc 154852
packaging
pkg of carton of 16
well surface area
4.0 cm2
working volume
1.0-2.0 mL
suitability
suitable for (cell culture; optimal for culturing fastidious cells (e.g. neurons))
General description
Nunc® Lab-Tek® II - CC2 Chamber Slide System
  • Chemically coated growth surface on glass slide that mimics polylysine
  • Provides binding sites optimal for fastidious cells (e.g. neurons)
  • Growth surface remains stable without refrigeration
  • Light blue frosted writing area
  • Slide seperator included in each pack
  • CE marked
Legal Information
CC2 is a trademark of Thermo Fisher Scientific or its subsidiaries
Chamber Slide is a trademark of Thermo Fisher Scientific or its subsidiaries
Lab-Tek is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Nunc is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Choose from one of the most recent versions:
Certificates of Analysis (COA)