S6045
Anti-Stem Cell Factor antibody produced in goat
IgG fraction of antiserum, lyophilized powder
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
biological source
goat
Quality Level
conjugate
unconjugated
antibody form
IgG fraction of antiserum
antibody product type
primary antibodies
clone
polyclonal
form
lyophilized powder
species reactivity
mouse
technique(s)
neutralization: suitable
storage temp.
−20°C
Gene Information
mouse ... Kitl(17311)
General description
Mast cells play an important role in innate immunity or the response of the body to invading pathogens. The functions of mast cells are regulated by a number of surface receptors; the receptors to Stem cell factor (SCF; c-Kit ligand or mast cell growth factor) being the most critical. The signalling triggered by SCF results in mast cell activation in response to antigens. The response involves tyrosine phosphorylation by specific tyrosine kinases that drive the downstream signalling process. The secondary pathways activated by SCF are signalling mediated by Akt, mTOR, MAPK and c-Jun. In addition to development of mast cells, SCF also regulates their subsequent maturation and homing to the residence sites. Constitutive activation by SCF is implicated in myeloproliferative discorders, systemic mastocytosis and gastrointestinal stomal cell tumors
Anti-Stem Cell Factor antibody reacts specifically with mouse stem cell factor.
Anti-Stem Cell Factor antibody reacts specifically with mouse stem cell factor.
Specificity
The antibody will neutralize the biological activity of recombinant mouse SCF.
Immunogen
recombinant mouse SCF
Application
Anti-Stem Cell Factor antibody may be used to neutralize SCF at a concentration of 25 ng/ml in 1X105/ml TF-1 cells. The antibody has been used to neutralize SCF in primary murine embryonic fibroblast (MEF) culture. The recommended concentration for indirect ELISA is 1.0 μg/ml and for immunoblotting is 2.0 μg/ml.
Physical form
Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline
Preparation Note
Purified using protein G
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Induction of mast cell proliferation, maturation, and heparin synthesis by the rat c-kit ligand, stem cell factor
Proceedings of the National Academy of Sciences of the USA, 15, 6382-6386 (1991)
Journal of leukocyte biology, 82(4), 915-925 (2007-07-27)
G-CSF and GM-CSF play important roles in regulating neutrophil production, survival, differentiation, and function. However, we have shown previously that G-CSF/GM-CSF double-deficient [knockout (KO)] mice still develop a profound neutrophilia in bone marrow and blood after infection with Candida albicans.
International archives of allergy and immunology, 107(1-3), 54-56 (1995-05-01)
Signaling through the c-kit receptor tyrosine kinase (Kit) is essential for development and survival of mast cells but not of basophils. Moreover, we recently found an activation mutation of Kit in several tumor mast cell lines.
The Journal of biological chemistry, 280(48), 40261-40270 (2005-09-24)
Stem cell factor (SCF) acts in synergy with antigen to enhance the calcium signal, degranulation, activation of transcription factors, and cytokine production in human mast cells. However, the underlying mechanisms for this synergy remain unclear. Here we show, utilizing bone
The Journal of allergy and clinical immunology, 117(6), 1214-1225 (2006-06-06)
The mast cell is a central player in allergy and asthma. Activation of these cells induces the release of preformed inflammatory mediators localized in specialized granules and the de novo synthesis and secretion of cytokines, chemokines, and eicosanoids. The balance
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service