Skip to Content
Merck
All Photos(1)

Key Documents

S4944

Sigma-Aldrich

Monoclonal Anti-Sonic Hedgehog (N-terminal) antibody produced in rat

clone 171018, purified immunoglobulin, lyophilized powder

Synonym(s):

Anti-Shh

Sign Into View Organizational & Contract Pricing

Select a Size

500 μG
€1,120.00

€1,120.00


Please contact Customer Service for Availability


Select a Size

Change View
500 μG
€1,120.00

About This Item

MDL number:
UNSPSC Code:
51111800
NACRES:
NA.41

€1,120.00


Please contact Customer Service for Availability

biological source

rat

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

171018, monoclonal

form

lyophilized powder

species reactivity

human, mouse

technique(s)

capture ELISA: suitable
immunohistochemistry (frozen sections): 25 μg/mL using mouse embryo
neutralization: suitable
western blot: 1-2 μg/test

isotype

IgG2a

UniProt accession no.

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SHH(6469)
mouse ... Shh(20423)

Related Categories

Immunogen

purified, E. coli-derived, recombinant mouse sonic hedgehog N-terminal peptide (amino acids 25-198).

Biochem/physiol Actions

Shh protein is secreted by Purkinje cells in the cerebellum with both autocrine and paracrine functions. The Shh signaling is mediated by the interaction of Shh with transmembrane proteins, Patched and Smoothened. The Shh pathway cross-talks Notch and Wnt pathways and acts via downstream targets such as cyclins, gamma catenin, myc and other transcription factors. The Hh family members have multiple roles as morphogens, cell survival factors, setting anterior-posterior axis and digit formation during development of limb and axon guidance factors during the development of brain and nervous system.

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline containing carbohydrates.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Raffaella Bonavita et al.
Biology open, 7(7) (2018-05-23)
In humans, the sacrum forms an important component of the pelvic arch, and it transfers the weight of the body to the lower limbs. The sacrum is formed by collapse of the intervertebral discs (IVDs) between the five sacral vertebrae
Chitra Lekha Dahia et al.
PloS one, 7(4), e35944-e35944 (2012-05-05)
Intervertebral discs (IVD) are essential components of the vertebral column. They maintain separation, and provide shock absorbing buffers, between adjacent vertebrae, while also allowing movements between them. Each IVD consists of a central semi-liquid nucleus pulposus (NP) surrounded by a
Alessia Omenetti et al.
American journal of physiology. Gastrointestinal and liver physiology, 294(3), G595-G598 (2008-01-26)
Hedgehog (Hh) signaling modulates tissue remodeling by controlling the fate of Hh-responsive cells. Healthy adult livers exhibit little Hh activity. However, cells involved in adult liver repair, including myofibroblasts and progenitors, are capable of producing and responding to Hh ligands.
Fergal C Kelleher
Carcinogenesis, 32(4), 445-451 (2010-12-28)
To conduct a systematic review of the role that the hedgehog signaling pathway has in pancreatic cancer tumorigenesis. PubMed search (2000-2010) and literature based references. Firstly, in 2009 a genetic analysis of pancreatic cancers found that a core set of
Shohreh F Farzan et al.
American journal of physiology. Gastrointestinal and liver physiology, 294(4), G844-G849 (2008-02-02)
The Hedgehog (Hh) family of secreted proteins is necessary for aspects of the development and maintenance of the gastrointestinal tract. Hh is thought to function as a morphogen, a mitogen, a cell survival factor, and an axon guidance factor. Given

Articles

Cancer stem cell media, spheroid plates and cancer stem cell markers to culture and characterize CSC populations.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service