Skip to Content
Merck
All Photos(1)

Documents

S4515

Sigma-Aldrich

Deamino-Tyr-β-Ala-secretin, pig

≥90% (HPLC)

Synonym(s):

DATA-Secretin, Desaminotyrosyl-β-alanylsecretin (pig), [Desaminotyrosyl-β-alanyl]-Secretin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C142H233N45O44
CAS Number:
Molecular Weight:
3274.64
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.26

Assay

≥90% (HPLC)

form

powder

composition

Peptide content, ~70%

technique(s)

radioimmunoassay: suitable

color

white

storage temp.

−20°C

SMILES string

CC(C)C[C@H](NC(=O)CNC(=O)[C@H](CCC(N)=O)NC(=O)[C@H](CC(C)C)NC(=O)[C@H](CC(C)C)NC(=O)[C@H](CCCNC(N)=N)NC(=O)[C@H](CCC(N)=O)NC(=O)[C@H](CC(C)C)NC(=O)[C@H](CCCNC(N)=N)NC(=O)[C@H](C)NC(=O)[C@H](CO)NC(=O)[C@H](CC(O)=O)NC(=O)[C@H](CCCNC(N)=N)NC(=O)[C@H](CC(C)C)NC(=O)[C@H](CCCNC(N)=N)NC(=O)[C@H](CO)NC(=O)[C@H](CC(C)C)NC(=O)[C@H](CCC(O)=O)NC(=O)[C@H](CO)NC(=O)[C@@H](NC(=O)[C@H](Cc1ccccc1)NC(=O)[C@@H](NC(=O)CNC(=O)[C@H](CC(O)=O)NC(=O)[C@H](CO)NC(=O)[C@H](Cc2c[nH]cn2)NC(=O)CCNC(=O)CCc3ccc(O)cc3)[C@@H](C)O)[C@@H](C)O)C(=O)N[C@@H](C(C)C)C(N)=O

InChI

1S/C142H233N45O44/c1-67(2)49-88(131(224)186-111(73(13)14)114(145)207)165-106(199)60-160-116(209)85(36-39-102(143)195)170-125(218)91(52-70(7)8)177-127(220)92(53-71(9)10)175-119(212)82(28-22-45-157-140(148)149)167-122(215)86(37-40-103(144)196)171-126(219)90(51-69(5)6)173-118(211)81(27-21-44-156-139(146)147)166-115(208)74(15)163-133(226)98(62-188)181-130(223)97(58-110(205)206)179-121(214)83(29-23-46-158-141(150)151)168-124(217)89(50-68(3)4)174-120(213)84(30-24-47-159-142(152)153)169-134(227)99(63-189)182-128(221)93(54-72(11)12)176-123(216)87(38-42-108(201)202)172-135(228)101(65-191)184-138(231)113(76(17)193)187-132(225)94(55-78-25-19-18-20-26-78)180-137(230)112(75(16)192)185-107(200)61-161-117(210)96(57-109(203)204)178-136(229)100(64-190)183-129(222)95(56-79-59-154-66-162-79)164-105(198)43-48-155-104(197)41-33-77-31-34-80(194)35-32-77/h18-20,25-26,31-32,34-35,59,66-76,81-101,111-113,188-194H,21-24,27-30,33,36-58,60-65H2,1-17H3,(H2,143,195)(H2,144,196)(H2,145,207)(H,154,162)(H,155,197)(H,160,209)(H,161,210)(H,163,226)(H,164,198)(H,165,199)(H,166,208)(H,167,215)(H,168,217)(H,169,227)(H,170,218)(H,171,219)(H,172,228)(H,173,211)(H,174,213)(H,175,212)(H,176,216)(H,177,220)(H,178,229)(H,179,214)(H,180,230)(H,181,223)(H,182,221)(H,183,222)(H,184,231)(H,185,200)(H,186,224)(H,187,225)(H,201,202)(H,203,204)(H,205,206)(H4,146,147,156)(H4,148,149,157)(H4,150,151,158)(H4,152,153,159)/t74-,75+,76+,81-,82-,83-,84-,85-,86-,87-,88-,89-,90-,91-,92-,93-,94-,95-,96-,97-,98-,99-,100-,101-,111-,112-,113-/m0/s1

InChI key

SSNOLQGOKGOOCG-DSOQYHOLSA-N

Looking for similar products? Visit Product Comparison Guide

Amino Acid Sequence

3-(4-Hydroxyphenyl)propionyl-β-Ala-His-Ser-Asp-Gly-Thr-Phe-Thr-Ser-Glu-Leu-Ser-Arg-Leu-Arg-Asp-Ser-Ala-Arg-Leu-Gln-Arg-Leu-Leu-Gln-Gly-Leu-Val-NH2

Application

Used as a tracer in a radioimmunoassay for secretin

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wunsch, E.
Hoppe-Seyler'S Zeitschrift fur Physiologische Chemie, 357, 1417-1417 (1976)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service