Skip to Content
Merck
All Photos(1)

Key Documents

S3922

Sigma-Aldrich

[Methyl-D-Phe6, D-Trp7,9, Leu10-psi(CH2NH)Leu11]-Substance P Fragment 6-11

≥97% (HPLC)

Synonym(s):

Methyl-D-Phe-D-Trp-Phe-D-Trp-Leu-psi[CH2NH]-Leu-NH2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C53H67N9O5
CAS Number:
Molecular Weight:
910.16
MDL number:
UNSPSC Code:
12352200

Assay

≥97% (HPLC)

composition

Peptide content, ~70%

UniProt accession no.

storage temp.

−20°C

SMILES string

CN[C@H](Cc1ccccc1)C(=O)N[C@H](Cc2c[nH]c3ccccc23)C(=O)N[C@@H](Cc4ccccc4)C(=O)N[C@H](Cc5c[nH]c6ccccc56)C(=O)N[C@H](CN[C@@H](CC(C)C)C(N)=O)CC(C)C

Gene Information

human ... TAC1(6863)

Biochem/physiol Actions

This substance P analog inhibited small-cell lung cancer proliferation in vitro and in vivo.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Orosz et al.
International journal of cancer, 60(1), 82-87 (1995-01-03)
Human small-cell lung-cancer cells (SCLC) produce and secrete gastrin-releasing peptide (GRP), the mammalian equivalent of bombesin (BN). There is some evidence to suggest that GRP is an autocrine regulator of SCLC cell growth. In the search for potent BN antagonists

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service