Skip to Content
Merck
All Photos(3)

Key Documents

S3007

Sigma-Aldrich

Sodium salicylate

ReagentPlus®, ≥99.5% (titration)

Synonym(s):

2-Hydroxybenzoic acid sodium salt, Salicylic acid sodium salt, Sodium 2-hydroxybenzoate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H4COONa
CAS Number:
Molecular Weight:
160.10
Beilstein:
3732792
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.25
Assay:
≥99.5% (titration)
form:
crystalline
solubility:
water: 575.7 g/L at 25 °C (77 °F)

Quality Level

product line

ReagentPlus®

Assay

≥99.5% (titration)

form

crystalline

color

white

useful pH range

6.85 (26.3 °C)

mp

>300 °C (lit.)

solubility

water: 575.7 g/L at 25 °C (77 °F)

SMILES string

[Na+].Oc1ccccc1C([O-])=O

InChI

1S/C7H6O3.Na/c8-6-4-2-1-3-5(6)7(9)10;/h1-4,8H,(H,9,10);/q;+1/p-1

InChI key

ABBQHOQBGMUPJH-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Sodium salicylate is the active ingredient in aspirin, commonly used as antipyretic, anti-inflammatory and analgesic drug. Sodium salicylate when taken in large quantities causes temporary hearing loss and tinnitus. Prolonged treatment with sodium salicylate reduces output from cochlea permanently, reduces compound action potential amplitude in vivo and neural degeneration in vitro. Sodium salicylate treatment in lateral amygdala alters tonography and excitability, causing hyperactivity in auditory cortex. Sodium salicylate impairs the expression of cellular-Finkel−Biskis−Jinkins murine osteogenic sarcoma virus (c-fos) and δ-FosB proteins in hippocampus region and affects spatial memory and learning. Sodium salicylate affects neurogenesis in dentate gyrus region of the brain.

Application

Sodium salicylate has been used:
  • to induce tinnitus, hyperacusis-like behavior in rats and infused directly into amygdala to measure local field potential.
  • to induce and confirm tinnitus by prepulse inhibition behavior (PPI) and its inhibition on learning and memory by Morris water maze test and subsequent gene expression studies in rats.
  • to enhance gamma amino butyric acid (GABA)ergic inhibition and enhance auditory response in the dorsal cortex of the inferior colliculus (ICd) measured by microiontophoretic technique.

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

208.9 °F - Pensky-Martens closed cup

Flash Point(C)

98.3 °C - Pensky-Martens closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effect of Long-Term Sodium Salicylate Administration on Learning, Memory, and Neurogenesis in the Rat Hippocampus
Niu H, et al.
BioMed Research International, 2018, 63-76 (2018)
Local application of sodium salicylate enhances auditory responses in the rat?s dorsal cortex of the inferior colliculus
Patel, Chirag R and Zhang, Huiming
Frontiers in Neurology, 5, 235-235 (2014)
Carmen S Padilla et al.
Frontiers in bioengineering and biotechnology, 8, 977-977 (2020-10-06)
Sugarcane and energy cane (Saccharum spp. hybrids) are ideal for plant-based production of recombinant proteins because their high resource-use efficiency, rapid growth and efficient photosynthesis enable extensive biomass production and protein accumulation at a cost-effective scale. Here, we aimed to
Raul Zavaliev et al.
Cell, 182(5), 1093-1108 (2020-08-19)
In plants, pathogen effector-triggered immunity (ETI) often leads to programmed cell death, which is restricted by NPR1, an activator of systemic acquired resistance. However, the biochemical activities of NPR1 enabling it to promote defense and restrict cell death remain unclear.
Josefa P Alameda et al.
Aging, 11(1), 127-159 (2019-01-12)
CYLD is a deubiquitinating enzyme known for its role as a tumor suppressor whose mutation leads to skin appendages tumors and other cancers. In this manuscript we report that the tumor suppressor CYLD, similarly to other renowned tumor suppressor genes

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service