Skip to Content
Merck
All Photos(1)

Key Documents

RAB1476

Sigma-Aldrich

Human GAPHD ELISA

for serum, plasma and cell culture supernatants

Sign Into View Organizational & Contract Pricing

Select a Size

96 WELLS
€831.00

€831.00


Please contact Customer Service for Availability


Select a Size

Change View
96 WELLS
€831.00

About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

€831.00


Please contact Customer Service for Availability

species reactivity

human

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable

input

sample type cell culture supernatant(s)
sample type serum
sample type plasma

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 0.82 ng/mL
standard curve range: 0.819-200 ng/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... GAPDH(2597)

General description

The GAPHD (glyceraldehyde-3-phosphate dehydrogenase) gene is mapped to human chromosome 12p13.31.[1] GAPHD encodes a multifunctional protein which is a glycolytic enzyme.[2][3]
This ELISA antibody pair detects human GAPHD. Other species not yet determined.

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached Protocolfor details.

Biochem/physiol Actions

GAPHD (glyceraldehyde-3-phosphate dehydrogenase) is associated with the progression of Huntington′s disease by increasing cytotoxicity.[4] Gaphd is often referred to as glycolytic enzyme and catalyzes the sixth step in glycolysis.[3][1] The Gaphd enzyme possess uracil-DNA glycosylase activity.[3] The enzyme regulates transcription, posttranscriptional gene expression, intracellular membrane trafficking and apoptosis.[2]

Other Notes

A sample Certificate of Analysis is available for this product. Please type the word sample in the text box provided for lot number.

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Glyceraldehyde-3-phosphate dehydrogenase (GAPDH) aggregation causes mitochondrial dysfunction during oxidative stress-induced cell death.
Nakajima H, et al.
The Journal of Biological Chemistry (2017)
A hydrocortisone derivative binds to GAPDH and reduces the toxicity of extracellular polyglutamine-containing aggregates.
Lazarev V F, et al.
Biochemical and Biophysical Research Communications, 487(3), 723-727 (2017)
Role of glyceraldehyde-3-phosphate dehydrogenase (GAPDH) in DNA repair
Kosova A A, et al.
Biochemistry (Moscow), 82(6), 643-654 (2017)
Prediction of potential cancer-risk regions based on transcriptome data: towards a comprehensive view.
Alisoltani A, et al.
PLoS ONE, 9(5), e96320-e96320 (2014)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service