Skip to Content
Merck
All Photos(1)

Key Documents

RAB1050

Sigma-Aldrich

Human Galectin-8 ELISA Kit

for cell culture supernatants, plasma, and serum samples

Sign Into View Organizational & Contract Pricing

Select a Size

96 WELLS
€528.00

€528.00


Please contact Customer Service for Availability


Select a Size

Change View
96 WELLS
€528.00

About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

€528.00


Please contact Customer Service for Availability

species reactivity

human

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable

input

sample type plasma
sample type serum sample(s)
sample type cell culture supernatant(s)

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 0.16 ng/ml

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... LGALS8(3964)

General description

This ELISA antibody pair detects Human Galectin-8 (LGALS8)

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached Protocolfor details.

Biochem/physiol Actions

Galectin-8 (Gal-8) is a member of “tandem-repeat” galectin family, possesses two carbohydrate binding domains linked by a short peptide. The short peptide accounts for Gal-8 cell adhesion and downstream signalling.[1] Tumor cells, as well as normal tissues, express Gal-8.[2] The expression level of Gal-8 is higher in multiple myeloma conditions and enhances adhesion to vascular endothelium.[1] Galectin-8 binds to integrin selectively and inhibits cell adhesion and promotes apoptosis.[3] Gal-8 interacts with integrin alpha and regulates neutrophil adhesion to endothelial cells and migration.[2] Gal-8 ligand binds to cluster of differentiation 44 (CD44) and causes an inflammatory response in rheumatoid arthritis.[4] Overexpression of Gal-8 plays a key role in metastasis of prostate cancer and cell survival by causing rearrangement of the cytoskeleton and E-cadherin expression.[5]

Other Notes

A sample Certificate of Analysis is available for this product. Please type the word sample in the text box provided for lot number.

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Galectin-8 modulates neutrophil function via interaction with integrin alphaM
Nishi N, et al.
Glycobiology, 13(11), 755-763 (2003)
Galectin-8 enhances adhesion of multiple myeloma cells to vascular endothelium and is an adverse prognostic factor
Friedel M, et al.
Glycobiology, 26(10), 1048-1058 (2016)
The involvement of CD44 and its novel ligand galectin-8 in apoptotic regulation of autoimmune inflammation
Sebban LE, et al.
Journal of Immunology, 179(2), 1225-1235 (2007)
Galectin-8 binding to integrins inhibits cell adhesion and induces apoptosis
Hadari YR, et al.
Journal of Cell Science, 113(13), 2385-2397 (2000)
Stable and high expression of Galectin-8 tightly controls metastatic progression of prostate cancer
Gentilini LD, et al.
Oncotarget, 8(27), 44654-44668 (2017)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service