Skip to Content
Merck
All Photos(1)

Key Documents

RAB0970

Sigma-Aldrich

Phosphotyrosine JAK1 / Tyrosine-protein Kinase JAK1 ELISA Kit

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
€528.00

€528.00


Please contact Customer Service for Availability


Select a Size

Change View
1 KIT
€528.00

About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

€528.00


Please contact Customer Service for Availability

species reactivity

human

technique(s)

ELISA: suitable

assay range

inter-assay cv: <10%
intra-assay cv: <12%

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... JAK1(3716)

General description

The antibody pair provided in this kit recognizes Human Tyrosine-Phosphorylated-JAK1.

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABHRP6Phosphotyrosine ELISA HRP-StreptavidinSDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service