Skip to Content
Merck
All Photos(2)

Key Documents

RAB0426

Sigma-Aldrich

Human P-Selectin ELISA Kit

for serum, plasma, cell culture supernatant and urine

Synonym(s):

Cytokine detection kit

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
€607.00

€607.00


Please contact Customer Service for Availability


Select a Size

Change View
1 KIT
€607.00

About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

€607.00


Please contact Customer Service for Availability

species reactivity

human

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable
capture ELISA: suitable

input

sample type plasma
sample type cell culture supernatant(s)
sample type urine
sample type serum

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 20 pg/mL
standard curve range: 0.041-30 ng/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... SELP(6403)

General description

The Human P-Selectin ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of human P-Selectin in serum, plasma, cell culture supernatants and urine.P-selectin, also known as cluster of differentiation 62 (CD62) or granule membrane protein 140 (GMP-140), is a Ca2+-dependent lectin. It is expressed within alpha-granules of platelets and the Weibel-Palade bodies of endothelial cells.[1]

Immunogen

Recombinant Human P-Selectin

Application

Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Human P-Selectin ELISA (Enzyme-Linked Immunosorbent Assay) kit has been used in ELISA to quantify P-selectin levels in platelet lysates.[2]
For research use only. Not for use in diagnostic procedures.

Biochem/physiol Actions

P-selectin serves as an adhesion molecule and plays an important role in leukocyte rolling and attachment.[1] In addition, it also acts a receptor for myeloid cells by associating with sialylated, fucosylated lactosaminoglycans.[3] Overexpression of P-selectin has been observed in apparently healthy women at risk for future vascular events.[1]

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABELADAELISA 1X Assay/Sample Diluent Buffer A (Item D1)SDS

  • RABELADBELISA 5X Assay/Sample Diluent Buffer B (Item E1)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Identification of a specific glycoprotein ligand for P-selectin (CD62) on myeloid cells.
Moore K L, et al.
The Journal of Cell Biology, 118(2), 445-456 (1992)
Soluble P-selectin and the risk of future cardiovascular events.
Ridker P M, et al.
Circulation, 103(4), 491-495 (2001)
Effects of selenium in the MAPK signaling cascade.
Rashtchizadeh N, et al.
Journal of cardiovascular and thoracic research, 7(3), 107-107 (2015)
Nadereh Rashtchizadeh et al.
Journal of cardiovascular and thoracic research, 7(3), 107-112 (2015-10-03)
This study aimed to discover by which mechanism selenium (Se) suppresses stimulated platelets stimulation in oxidative stress underlying diseases. Human platelets pretreated with Se and stimulated by Cu(2+)-oxidized low density of lipoprotein (OxLDL) or thrombin before assessment of P-selectin and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service