Skip to Content
Merck
All Photos(2)

Key Documents

RAB0259

Sigma-Aldrich

Human IL-15 ELISA Kit

for serum, plasma, cell culture supernatant and urine

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
€528.00

€528.00


Please contact Customer Service for Availability


Select a Size

Change View
1 KIT
€528.00

About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

€528.00


Please contact Customer Service for Availability

species reactivity

human

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable
capture ELISA: suitable

input

sample type urine
sample type plasma
sample type serum
sample type cell culture supernatant(s)

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 3 pg/mL
standard curve range: 3.3-800 pg/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... IL15(3600)

General description

Interleukin-15 (IL-15) is a four α-helical bundle cytokine[1] expressed in the immune system as well as in the skeletal muscles.[2] The IL-15 gene is mapped to human chromosome 4q31.21.[3]
The Human IL-15 ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of human IL-15 in serum, plasma, cell culture supernatants and urine.

Immunogen

Recombinant Human IL-15

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Biochem/physiol Actions

Interleukin-15 (IL-15) is an immunomodulatory cytokine,[2] that plays a key role in the cluster of differentiation CD8+ T cell memory..[2] It plays a crucial role in the healing of chronic wounds.[1] IL -15 has been implicated in the pathogenesis of autoimmune diseases.[4]

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABELADAELISA 1X Assay/Sample Diluent Buffer A (Item D1)SDS

  • RABELADBELISA 5X Assay/Sample Diluent Buffer B (Item E1)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Krzysztof Ficek et al.
Frontiers in physiology, 10, 876-876 (2019-07-25)
Based on the important role of interleukin-15 (IL-15) in human metabolism and, in consequence, in body composition modulation, we examined whether rs1589241 and rs1057972 polymorphisms, analyzed individually or in combination, would influence the effects of a training program. Accordingly, we
A M Jones et al.
International journal of molecular medicine, 38(3), 679-686 (2016-07-28)
Chronic wounds represent a significant burden to health services and are associated with patient morbidity. Novel methods to diagnose and/or treat problematic wounds are needed. Interleukin (IL)-15 is a cytokine involved in a number of biological processes and disease states such
Wei Fang et al.
Comparative biochemistry and physiology. Part B, Biochemistry & molecular biology, 143(3), 335-343 (2006-02-04)
Interleukin-15 (IL-15) plays an important role in adaptive immune systems in vertebrates with similar bioactivities to interleukin-2 (IL-2). Here we report molecular cloning, sequence analysis and distribution of an IL-15 homologue from a pufferfish (Tetraodon nigroviridis). It is located within
Thomas A Waldmann
Arthritis research & therapy, 6(4), 174-177 (2004-07-01)
Interleukin (IL)-15 is a dangerous inflammatory cytokine that induces tumor-necrosis factor-alpha, IL-1beta and inflammatory chemokines. It inhibits self-tolerance mediated by IL-2 mediated activation-induced cell death and facilitates maintenance of CD8+ memory T-cell survival including that of self-directed memory cells. Disordered
Pierfrancesco Marra et al.
Cancer research, 74(17), 4908-4921 (2014-07-02)
Despite its aggressive nature, triple-negative breast cancer (TNBC) often exhibits leucocyte infiltrations that correlate with favorable prognosis. In this study, we offer an explanation for this apparent conundrum by defining TNBC cell subsets that overexpress the IL15 immune receptor IL15RA.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service