Skip to Content
Merck
All Photos(2)

Key Documents

RAB0214

Sigma-Aldrich

Mouse HGF ELISA Kit

for serum, plasma and cell culture supernatant

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
€528.00

€528.00


Please contact Customer Service for Availability


Select a Size

Change View
1 KIT
€528.00

About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

€528.00


Please contact Customer Service for Availability

species reactivity

mouse

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable
capture ELISA: suitable

input

sample type plasma
sample type serum
sample type cell culture supernatant(s)

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 400 pg/mL
standard curve range: 0.614-60 ng/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

mouse ... Hgf(15234)

General description

The Mouse HGF (Hepatocyte Growth Factor) ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of mouse HGF in serum, plasma, cell culture supernatants and urine.

Immunogen

Recombinant Mouse HGF

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABELADAELISA 1X Assay/Sample Diluent Buffer A (Item D1)SDS

  • RABELADBELISA 5X Assay/Sample Diluent Buffer B (Item E1)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Caroline A Enns et al.
Blood, 136(8), 989-1001 (2020-05-10)
Matriptase-2 (MT2), encoded by TMPRSS6, is a membrane-anchored serine protease that plays a key role in suppressing hepatic hepcidin expression. MT2 is synthesized as a zymogen and undergoes autocleavage for activation. Previous studies suggest that MT2 suppresses hepcidin by cleaving
Jesse Hudson et al.
Molecular and cellular biology, 34(19), 3689-3701 (2014-07-30)
Breast cancers are stratified into distinct subtypes, which influence therapeutic responsiveness and patient outcome. Patients with luminal breast cancers are often associated with a better prognosis relative to that with other subtypes. However, subsets of patients with luminal disease remain
Ji-Dong Chen et al.
Asian Pacific journal of cancer prevention : APJCP, 15(11), 4545-4548 (2014-06-28)
To investigate the changes of serum vascular endothelial growth factor (VEGF), soluble interleukin-2 receptor (SIL-2R) and hepatocyte growth factor (HGF) contents in patients with primary hepatocellular carcinoma (HCC) before and after percutaneous microwave coagulation therapy (PMCT) and determine their clinical
Elisa Tinazzi et al.
Immunology letters, 161(2), 236-240 (2014-01-23)
CD30 and CD30 ligand (CD30L) are members of TNF-receptor and TNF superfamilies respectively. CD30(+)T cells are increased in several diseases and interaction between CD30(+) and CD30L(+)T cells leads either to cell proliferation or apoptosis. In patients with rheumatoid arthritis (RA)
Mishaal B Patel et al.
Carcinogenesis, 35(8), 1891-1900 (2014-05-31)
Activated cancer-associated human pancreatic stellate cells (CAhPSCs, which produce the collagenous stroma of pancreatic cancer [PC]) are known to play a major role in PC progression. Apart from inducing cancer cell proliferation and migration, CAhPSCs have also been implicated in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service