Skip to Content
Merck
All Photos(1)

Key Documents

RA462250-EP

Rotor for Eppendorf® 5804/5804R & 5810/5810R Centrifuges

A-4-62, for 5810/5810R, aerosol-tight, with 4 × 250 mL rectangular buckets

Synonym(s):

Lid for Eppendorf® FA-45-48-11 Rotor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41103909
Product RA462250-EP is not currently sold in your country. Contact Technical Service

feature

to hold 0.2 mlLto 250 mL tubes and bottles
aerosol-tight
economic swing- bucket rotor
for 5810/5810R
with 4 × 250 mL rectangular buckets

packaging

pack of 1 ea

manufacturer/tradename

Eppendorf® 022638009 (NA)
Eppendorf® 5810709008 (EU)

parameter

3,200 × g max. RCF

technique(s)

centrifugation: suitable

compatibility

for use with centrifuge models 5810/5810R

Looking for similar products? Visit Product Comparison Guide

General description

A wide selections of rotors to fit the Eppendorf 5804 and 5810 centrifuges.

Features and Benefits

  • The swing-bucket rotor has a maximum speed of 3,220 × g (4,000 rpm).
  • Rotor A-4-62 is designed to accommodate tubes, bottles ranging from 0.2 mL to 250 mL, as well as plates.
  • For enhanced safety during the centrifugation of hazardous samples, aerosol-tight caps are available as an optional feature.

Legal Information

Eppendorf is a registered trademark of Eppendorf AG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service