Skip to Content
Merck
All Photos(1)

Key Documents

R5648

Sigma-Aldrich

Rottlerin

Synonym(s):

1-[6-[(3-Acetyl-2,4,6-trihydroxy-5-methylphenyl)methyl]-5,7-dihydroxy-2,2-dimethyl-2H-1-benzopyran-8-yl]-3-phenyl-2-propen-1-one, Mallotoxin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H28O8
CAS Number:
Molecular Weight:
516.54
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

biological source

plant (Mallotus philippinensis)

Assay

≥95% (HPLC)

form

powder

mp

200.0 °C

solubility

ethanol: 1 mg/mL

storage temp.

2-8°C

SMILES string

CC(=O)c1c(O)c(C)c(O)c(Cc2c(O)c3C=CC(C)(C)Oc3c(c2O)C(=O)\C=C\c4ccccc4)c1O

InChI

1S/C30H28O8/c1-15-24(33)19(27(36)22(16(2)31)25(15)34)14-20-26(35)18-12-13-30(3,4)38-29(18)23(28(20)37)21(32)11-10-17-8-6-5-7-9-17/h5-13,33-37H,14H2,1-4H3/b11-10+

InChI key

DEZFNHCVIZBHBI-ZHACJKMWSA-N

Gene Information

human ... CDK2(1017)

Looking for similar products? Visit Product Comparison Guide

General description

Rottlerin is extracted from Mallotus philippinensis. It is a protein kinase Cδ (PKCδ) inhibitor. Rottlerin acts as an uncoupler of mitochondrial respiration from oxidative phosphorylation. It has antitumor, autophagy, anti-proliferative, anti-metastasis and anti-invasive properties.

Application

Rottlerin has been used:
  • as a protein kinase C δ (PKCδ) inhibitor for the analysis of protein content and phosphorylation in rats
  • to examine the gene expression of early osteoblast markers
  • to attenuate pyruvate dehydrogenase (PDH) phosphorylation by succinate
  • to depolarise mitochondria
  • to investigate its antitumor activity in pancreatic cancer (PC) cells

Biochem/physiol Actions

Potent activator of the large conductance voltage and Ca2+ activated K+ channel; hERG channel activator.
Recently, Rottlerin (mallotoxin) has been shown to be a potent activator of the large conductance voltage and Ca2 activated K+ channel and to potently leftward shift the conductance-voltage relationship of the channel. Mallatoxin tested on hERG channels increased both step and tail hERG current by leftward shifting the voltage dependence of hERG activation and slowing channel deactivation. These actions distinguish Mallatoxin as a novel naturally occurring hERG channel activator.

Features and Benefits

This compound is featured on the PKC page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rottlerin exhibits anti-cancer effect through inactivation of S phase kinase-associated protein 2 in pancreatic cancer cells
Su J, et al.
American Journal of Cancer Research, 6(10), 2178-2178 (2016)
Stephen P Soltoff
Trends in pharmacological sciences, 28(9), 453-458 (2007-08-19)
Rottlerin has been used as a protein kinase Cdelta (PKCdelta)-selective inhibitor in hundreds of studies, on the basis of initial substrate phosphorylation studies in vitro. However, in more recent studies, rottlerin did not block PKCdelta activity but did block other
Hyperammonemia alters membrane expression of GluA1 and GluA2 subunits of AMPA receptors in hippocampus by enhancing activation of the IL-1 receptor: underlying mechanisms
Taoro-Gonzalez L, et al.
Journal of Neuroinflammation, 15(1), 36-36 (2018)
Brahma N Singh et al.
Biochemical pharmacology, 84(9), 1154-1163 (2012-08-21)
Multiple lines of evidence support the idea that autophagy plays an essential role in the development of drug resistance, self-renewal, differentiation, and tumorigenic potentials of cancer stem cells (CSCs). Rottlerin (ROT) is widely used as a protein kinase C-delta (PKC-δ)
Rottlerin exhibits antitumor activity via down-regulation of TAZ in non-small cell lung cancer
Zhao Z, et al.
Testing, 8(5), 7827-7827 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service