Skip to Content
Merck
All Photos(2)

Key Documents

R3629

Sigma-Aldrich

Ribonucleic acid diethylaminoethanol salt

Type IX

Synonym(s):

Ribonucleic acid from torula yeast, RNA

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€277.00
25 G
€1,030.00

€277.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€277.00
25 G
€1,030.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
41106305
NACRES:
NA.51

€277.00


Please contact Customer Service for Availability

Request a Bulk Order

type

Type IX

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Ribonucleic acid (RNA) from torula yeast may be used as a substrate for studying ribonuclease activities of enzymes such as ribonuclease-A, ribonuclease T1 (RNAase) and bougainvillea xbuttiana antiviral protein 1 (BBAP1).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yvonne Tay et al.
Nature, 505(7483), 344-352 (2014-01-17)
Recent reports have described an intricate interplay among diverse RNA species, including protein-coding messenger RNAs and non-coding RNAs such as long non-coding RNAs, pseudogenes and circular RNAs. These RNA transcripts act as competing endogenous RNAs (ceRNAs) or natural microRNA sponges
Ricardo Saldaña-Meyer et al.
Genes & development, 28(7), 723-734 (2014-04-04)
The multifunctional CCCTC-binding factor (CTCF) protein exhibits a broad range of functions, including that of insulator and higher-order chromatin organizer. We found that CTCF comprises a previously unrecognized region that is necessary and sufficient to bind RNA (RNA-binding region [RBR])
Hilal Kazan et al.
Nucleic acids research, 41(Web Server issue), W180-W186 (2013-06-12)
RBPmotif web server (http://www.rnamotif.org) implements tools to identify binding preferences of RNA-binding proteins (RBPs). Given a set of sequences that are known to be bound and unbound by the RBP of interest, RBPmotif provides two types of analysis: (i) de
Guo-Liang Chew et al.
Development (Cambridge, England), 140(13), 2828-2834 (2013-05-24)
Large-scale genomics and computational approaches have identified thousands of putative long non-coding RNAs (lncRNAs). It has been controversial, however, as to what fraction of these RNAs is truly non-coding. Here, we combine ribosome profiling with a machine-learning approach to validate
Giulia Biffi et al.
Nature chemistry, 6(1), 75-80 (2013-12-19)
Following extensive evidence for the formation of four-stranded DNA G-quadruplex structures in vitro, DNA G-quadruplexes have been observed within human cells. Although chemically distinct, RNA can also fold in vitro into G-quadruplex structures that are highly stable because of the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service