Skip to Content
Merck
All Photos(3)

Documents

R2779

Sigma-Aldrich

Renin human

BioUltra, recombinant, expressed in mammalian cells, ≥90 unit/μg protein, ≥98% (SDS-PAGE)

Sign Into View Organizational & Contract Pricing


About This Item

Enzyme Commission number:
MDL number:
UNSPSC Code:
12352204

recombinant

expressed in mammalian cells

product line

BioUltra

Assay

≥98% (SDS-PAGE)

specific activity

≥90 unit/μg protein

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... REN(5972)

Application

Human renin is a member of the Peptidase A1 family. It catalyzes the hydrolysis of angiotensinogen to produce angiotensin I. It is used to study human renin-dependent hypertension as well as renin induced obesity .

Biochem/physiol Actions

Renin is a 406 amino acid precursor (45,057 Da) containing a 23 amino acid signal peptide and a 43 amino acid propeptide. Native active renin is a 340 amino acid glycoprotein containing three disulfide bonds. Renin, an aspartyl protease, cleaves angiotensinogen which results in Ang I. Ang I is converted to Ang II by the Ang-converting enzyme (ACE) . Prorenin is hydrolyzed by plasma kallekrein to produce active renin. The human renin transgene is thought to be responsible for increased appetite, obesity, and metabolic changes which are not related to angiotensin II .

Physical properties

Renin is processed as a 406 amino acid precursor (45,057 Da) containing a 23 amino acid signal peptide and a 43 amino acid propeptide. Native active renin is a 340 amino acid glycoprotein containing three disulfide bonds.
Supplied as a solution in 50 mM Tris HCl, pH 8

Unit Definition

One unit will produce one fluorescent unit from the hydrolysis of Renin Substrate I (Arg-Glu(EDANS)-Ile-His-Pro-Phe-His-Leu-Val-Ile-His-Thr-Lys(dabcyl)-Arg) at 37 °C at pH 8.0

Preparation Note

Expressed in mammalian cells as prorenin. The prorenin is activated to renin using trypsin and re-purified.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Manne Krop et al.
Hypertension (Dallas, Tex. : 1979), 61(5), 1075-1082 (2013-03-06)
Renin inhibitors like aliskiren not only block renin but also bind prorenin, thereby inducing a conformational change (like the change induced by acid) allowing its recognition in a renin-specific assay. Consequently, aliskiren can be used to measure prorenin. VTP-27999 is
Cameron J Gilbert et al.
The Canadian journal of cardiology, 29(5), 586-591 (2013-04-02)
Some evidence suggests that the direct renin inhibitor aliskiren may increase the risk of severe hyperkalemia, stroke, or acute kidney injury (AKI) when prescribed with angiotensin-converting enzyme inhibitors (ACEi's) or angiotensin-receptor blockers (ARBs). The extent to which concomitant treatment increases
Stephen J Nicholls et al.
JAMA, 310(11), 1135-1144 (2013-09-04)
Blood pressure reduction and renin-angiotensin-aldosterone system inhibition are targets for treatment of atherosclerosis. The effect of renin inhibition on coronary disease progression has not been investigated. To determine the effects of renin inhibition with aliskiren on progression of coronary atherosclerosis.
Brian G Keevil
Best practice & research. Clinical endocrinology & metabolism, 27(5), 663-674 (2013-10-08)
Liquid chromatography tandem mass spectrometry (LC-MS/MS) is increasingly becoming the method of choice for steroid hormone measurements due to small sample volumes, fast analysis times and improved specificity compared to immunoassays. Achievement of demanding analytical targets for steroid analysis is
Mihai Gheorghiade et al.
JAMA, 309(11), 1125-1135 (2013-03-13)
Hospitalizations for heart failure (HHF) represent a major health burden, with high rates of early postdischarge rehospitalization and mortality. To investigate whether aliskiren, a direct renin inhibitor, when added to standard therapy, would reduce the rate of cardiovascular (CV) death

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service