R1276
Reversin 121
≥97% (HPLC), powder
Synonym(s):
Boc-Asp(Obzl)-Lys(Z)-OtBu
Select a Size
€414.00
Select a Size
About This Item
€414.00
Recommended Products
Quality Level
Assay
≥97% (HPLC)
form
powder
color
white
mp
200 °C
solubility
DMSO or ethanol: soluble
storage temp.
−20°C
SMILES string
CC(C)(C)OC(=O)N[C@@H](CC(=O)OCc1ccccc1)C(=O)N[C@@H](CCCCNC(=O)OCc2ccccc2)C(=O)OC(C)(C)C
InChI
1S/C34H47N3O9/c1-33(2,3)45-30(40)26(19-13-14-20-35-31(41)44-23-25-17-11-8-12-18-25)36-29(39)27(37-32(42)46-34(4,5)6)21-28(38)43-22-24-15-9-7-10-16-24/h7-12,15-18,26-27H,13-14,19-23H2,1-6H3,(H,35,41)(H,36,39)(H,37,42)/t26-,27-/m0/s1
InChI key
SVNKEDMVAQBLLN-SVBPBHIXSA-N
Gene Information
human ... GRIN2A(2903)
mouse ... GRIN2A(14811)
rat ... GRIN2A(24409)
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Protein-based drug transporters are expressed in Sf9 cells. Understanding the specific mechanisms of tumor cell transporters is an essential aspect of chemotherapeutic drug design.
Cancer stem cell media, spheroid plates and cancer stem cell markers to culture and characterize CSC populations.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service