Recommended Products
biological source
rat skin (Sprague Dawley)
Quality Level
packaging
pkg of 500,000 cells
manufacturer/tradename
Cell Applications, Inc
growth mode
Adherent
morphology
Epidermal
technique(s)
cell culture | mammalian: suitable
relevant disease(s)
allergies; cancer
shipped in
dry ice
storage temp.
−196°C
General description
REK from Cell Applications, Inc. provide a suitable model system to study many aspects of epithelial function and disease, particularly those related to skin biology and toxicology. It was shown that TGFβ1 treatment decreased c-Myc mRNA in REK which, taken together with the TGFβ1-induced growth arrest exhibited by keratinocytes and opposite results obtained in other cells, suggests a correlation between c-Myc mRNA expression and the mitogenic response (Nakai, 2008). Additionally, REK were used to demonstrate that pathological increases in keratinocyte sodium channels Nav1.1, Nav1.6, and Nav1.8 expression may contribute to pain by increasing epidermal ATP release, resulting in excessive activation of P2X receptors on primary sensory axons (Zhao, 2008). REK were also used to demonstrate that laminin 332 deposition is inhibited by ionizing radiation and, in combination with slower keratinocyte migration, can contribute to the delayed wound healing of irradiated skin (Jourdan, 2011).
Cell Line Origin
Application
Components
Quality
Preparation Note
- Primary culture, >500,000 cells in Rat Keratinocyte Basal Medium that contains 10% FBS and 10% DMSO
- These epithelial cells proliferate well when thawed and plated from cryopreservation, but they do not plate and proliferate after trypsinization
Subculture Routine
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service