Skip to Content
Merck
All Photos(1)

Key Documents

Q2001

Sigma-Aldrich

Quinoline

~98%, liquid

Synonym(s):

1-Benzazine, 2,3-Benzopyridine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H7N
CAS Number:
Molecular Weight:
129.16
Beilstein:
107477
EC Number:
MDL number:
UNSPSC Code:
12352103

vapor density

4.5 (vs air)

vapor pressure

0.07 mmHg ( 20 °C)

Assay

~98%

form

liquid

autoignition temp.

896 °F

color

clear brown

refractive index

n20/D 1.625 (lit.)

bp

113-114 °C/11 mmHg (lit.)
237 °C (lit.)

mp

−17-−13 °C (lit.)

density

1.093 g/mL at 25 °C (lit.)

SMILES string

c1ccc2ncccc2c1

InChI

1S/C9H7N/c1-2-6-9-8(4-1)5-3-7-10-9/h1-7H

InChI key

SMWDFEZZVXVKRB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Aquatic Chronic 2 - Carc. 1B - Eye Irrit. 2 - Muta. 2 - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

213.8 °F - closed cup

Flash Point(C)

101 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fang-Fang Wang et al.
Organic & biomolecular chemistry, 10(43), 8605-8608 (2012-10-09)
A practical Brønsted acid promoted benzylic C-H functionalization of 2-methylazaarenes and nucleophilic addition to aldehydes was developed in good to excellent yields. A six-membered hydrogen-bonding transition state is proposed to be crucial for the reaction. Ready availability of the two
Jin Hoon Kim et al.
Dalton transactions (Cambridge, England : 2003), 42(15), 5500-5507 (2013-02-22)
Zinc sensors based on 2,3-dipicolylamine (DPA) and quinoline have been synthesized. They fluoresced in the presence of Zn(2+) and remained fluorescent when other metal ions were present. Fluorescence enhancement of the sensors was not seen for most other metal ions.
Yu Zhang et al.
Dalton transactions (Cambridge, England : 2003), 41(38), 11776-11782 (2012-08-21)
A series of carboxamidoquinoline-based fluorescent sensors (the AQZ family) were synthesized and characterized. The AQZ family members were highly soluble in water and showed good selectivity for Zn(2+)via enhanced fluorescence in aqueous buffer solution. Fluorescence signals could be tuned from
Narendra Reddy Chereddy et al.
Dalton transactions (Cambridge, England : 2003), 41(38), 11753-11759 (2012-08-21)
Two new rhodamine based probes 1 and 2 for the detection of Fe(3+) were synthesized and their selectivity towards Fe(3+) ions in the presence of other competitive metal ions tested. The probe 1 formed a coloured complex with Fe(3+) as
Sergey Kapishnikov et al.
Proceedings of the National Academy of Sciences of the United States of America, 109(28), 11188-11193 (2012-06-30)
Heme detoxification is a critical step in the life cycle of malaria-causing parasites, achieved by crystallization into physiologically insoluble hemozoin. The mode of nucleation has profound implications for understanding the mechanism of action of antimalarial drugs that inhibit hemozoin growth.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service