Skip to Content
Merck
All Photos(1)

Key Documents

Q1626

Sigma-Aldrich

Quene 1-AM

Synonym(s):

8-bis(Carboxymethyl)amino-2-[2-(2-bis(carboxymethyl)aminophenyl)ethenyl]-6-methoxyquinoline tetra(acetoxymethyl) ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C38H41N3O17
CAS Number:
Molecular Weight:
811.74
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

shipped in

dry ice

storage temp.

−20°C

SMILES string

COc1cc(N(CC(=O)OCOC(C)=O)CC(=O)OCOC(C)=O)c2nc(\C=C\c3ccccc3N(CC(=O)OCOC(C)=O)CC(=O)OCOC(C)=O)ccc2c1

InChI

1S/C38H41N3O17/c1-24(42)51-20-55-34(46)16-40(17-35(47)56-21-52-25(2)43)32-9-7-6-8-28(32)10-12-30-13-11-29-14-31(50-5)15-33(38(29)39-30)41(18-36(48)57-22-53-26(3)44)19-37(49)58-23-54-27(4)45/h6-15H,16-23H2,1-5H3/b12-10+

InChI key

LEFGNZVSFRGPIG-ZRDIBKRKSA-N

Application

Fluorescent probe for intracellular pH.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Rogers et al.
The Journal of biological chemistry, 258(10), 5994-5997 (1983-05-25)
A new fluorescent intracellular pH indicator is described ("quene 1") which is related to the tetracarboxylate Ca2+ indicator based on the quinoline fluorophor ("quin 2"). Quene 1 has excitation and emission maxima at 390 and 530 nm, respectively, and shows
J Rogers et al.
FEBS letters, 161(1), 21-27 (1983-09-05)
Photometric fluorescence microscopy has been used to measure intracellular pH (pHi) and free calcium concentrations [( Ca]i) in individual mouse thymocytes and 2H3 rat basophil leukaemic cells containing indicators for pH (quene 1) or calcium (quin 2). The pHi and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service