Skip to Content
Merck
All Photos(1)

Key Documents

PZ0391

Sigma-Aldrich

PF-06745268

≥98% (HPLC)

Synonym(s):

2-[[(1aS,7bS)-1a,2-Dihydro-2,2-dimethyl-6-(trifluoromethyl)benzo[b]cyclopropa[d]pyran-7b(1H)-yl]methyl]-3,4-dihydro-7-(4-methyl-1H-imidazol-1-yl)-2H-pyrido[1,2-a]pyrazine-1,6-dione

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€64.40

€64.40


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€64.40

About This Item

Empirical Formula (Hill Notation):
C26H25F3N4O3
CAS Number:
Molecular Weight:
498.50
UNSPSC Code:
51111800
NACRES:
NA.77

€64.40


Please contact Customer Service for Availability

Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

optical activity

[α]/D -13 to -17°, c = 1.0 in methanol

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C1C(N2C=C(N=C2)C)=CC=C3N1CCN(C[C@@]4(C5)[C@@]5([H])C(C)(C)OC6=C4C=C(C(F)(F)F)C=C6)C3=O

InChI key

LHTSLMHEFKNOOX-PXDATVDWSA-N

Biochem/physiol Actions

PF-06745268 is an orally available, brain penetrant, potent and selective γ-secretase modulator. PF-06745268 induces robust and sustained reduction of brain amyloid-β42 (Aβ42) in rats.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service