Skip to Content
Merck
All Photos(1)

Key Documents

PZ0374

Sigma-Aldrich

PF-06446846 hydrochloride

≥98% (HPLC)

Synonym(s):

N-(3-Chloropyridin-2-yl)-N-((3R)-piperidin-3-yl)-4-(3H-[1,2,3]triazolo[4,5-b]pyridin-3-yl)benzamide hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€144.00
25 MG
€574.00

€144.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€144.00
25 MG
€574.00

About This Item

Empirical Formula (Hill Notation):
C22H20ClN7O · xHCl
CAS Number:
Molecular Weight:
433.89 (free base basis)
UNSPSC Code:
41121800
NACRES:
NA.77

€144.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

optical activity

[α]/D -60 to -50°, c = 1 in methanol

storage condition

desiccated

color

white to beige

solubility

H2O: 2 mg/mL, clear

storage temp.

room temp

SMILES string

O=C(C1=CC=C(N2C(N=CC=C3)=C3N=N2)C=C1)N([C@H]4CNCCC4)C5=C(Cl)C=CC=N5

InChI

1S/C22H20ClN7O/c23-18-5-2-12-25-20(18)29(17-4-1-11-24-14-17)22(31)15-7-9-16(10-8-15)30-21-19(27-28-30)6-3-13-26-21/h2-3,5-10,12-13,17,24H,1,4,11,14H2/t17-/m1/s1

InChI key

FDTXHWQFIXYHCL-QGZVFWFLSA-N

Biochem/physiol Actions

PF-06446846 is a selective orally active PCSK9 inhibitor that appears to act by causing the ribosome to stall when synthesizing PCSK9. PCSK9 binds to the LDL receptor, preventing it from removing LDL cholesterol from the blood. PF-06446846 was found to reduce plasma PCSK9 and total cholesterol levels in rats.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nathanael G Lintner et al.
PLoS biology, 15(3), e2001882-e2001882 (2017-03-23)
Proprotein convertase subtilisin/kexin type 9 (PCSK9) plays a key role in regulating the levels of plasma low-density lipoprotein cholesterol (LDL-C). Here, we demonstrate that the compound PF-06446846 inhibits translation of PCSK9 by inducing the ribosome to stall around codon 34

Questions

  1. Re: PZ0374 (PF-06446846 hydrochloride) I see that it is recommended to resuspend in H2O at 2 mg/mL. Would this solution then be stored at -20 degrees? How long is it stable in this format? Thank you, Maya

    1 answer
    1. The stability of this product in solution has not been determined.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service