Skip to Content
Merck
All Photos(1)

Key Documents

PZ0334

Sigma-Aldrich

PF-06422913

≥98% (HPLC)

Synonym(s):

1-Methyl-6-(2-pyridinylmethoxy)-3-[4-(trifluoromethyl)-3-pyridinyl]-1H-pyrazolo[3,4-b]prazine, PF06422913

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€61.20
25 MG
€248.00

€61.20


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€61.20
25 MG
€248.00

About This Item

Empirical Formula (Hill Notation):
C18H13F3N6O
CAS Number:
Molecular Weight:
386.33
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

€61.20


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 10 mg/mL, clear

storage temp.

room temp

SMILES string

CN1N=C(C2=C(C(F)(F)F)C=CN=C2)C3=NC=C(OCC4=CC=CC=N4)N=C31

InChI

1S/C18H13F3N6O/c1-27-17-16(15(26-27)12-8-22-7-5-13(12)18(19,20)21)24-9-14(25-17)28-10-11-4-2-3-6-23-11/h2-9H,10H2,1H3

InChI key

UHUFBNVEMLJTKS-UHFFFAOYSA-N

Biochem/physiol Actions

PF-06422913 is an orally active, potent and selective metabotropic glutamate receptor 5 (mGluR5) negative allosteric modulator.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service