Skip to Content
Merck
All Photos(1)

Key Documents

PZ0324

Sigma-Aldrich

PF-06263276

≥98% (HPLC)

Synonym(s):

PF06263276, [2-[6-(2-Ethyl-5-fluoro-4-hydroxyphenyl)-1H-indazol-3-yl]-3,4,6,7-tetrahydro-5H-imidazo[4,5-c]pyridin-5-yl][5-(1-piperidinyl)-2-pyrazinyl]-methanone

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€87.60
25 MG
€353.00

€87.60


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€87.60
25 MG
€353.00

About This Item

Empirical Formula (Hill Notation):
C31H31FN8O2
CAS Number:
Molecular Weight:
566.63
UNSPSC Code:
12352200
NACRES:
NA.77

€87.60


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 5 mg/mL, clear

storage temp.

room temp

SMILES string

FC1=C(O)C=C(CC)C(C2=CC(NN=C3C4=NC5=C(CCN(C(C6=CN=C(N7CCCCC7)C=N6)=O)C5)N4)=C3C=C2)=C1

InChI

1S/C31H31FN8O2/c1-2-18-13-27(41)22(32)14-21(18)19-6-7-20-24(12-19)37-38-29(20)30-35-23-8-11-40(17-26(23)36-30)31(42)25-15-34-28(16-33-25)39-9-4-3-5-10-39/h6-7,12-16,41H,2-5,8-11,17H2,1H3,(H,35,36)(H,37,38)

InChI key

XDJGNPSZQSWJCV-UHFFFAOYSA-N

Biochem/physiol Actions

PF-06263276 is an inhibitor of the Janus kinase (JAK) enzymes 1, 2, 3 and tyrosine kinase 2 (TYK2). PF-06263276 has been investigated for the treatment of psoriasis.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Peter Jones et al.
Journal of medicinal chemistry, 60(2), 767-786 (2016-12-17)
By use of a structure-based computational method for identification of structurally novel Janus kinase (JAK) inhibitors predicted to bind beyond the ATP binding site, a potent series of indazoles was identified as selective pan-JAK inhibitors with a type 1.5 binding

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service