Skip to Content
Merck
All Photos(1)

Key Documents

PZ0294

Sigma-Aldrich

PF-04880594

≥98% (HPLC)

Synonym(s):

3-[[4-[1-(2,2-Difluoroethyl)-3-(1H-pyrrolo[2,3-b]pyridin-5-yl)-1H-pyrazol-4-yl]-2-pyrimidinyl]amino]-propanenitrile, PF 04880594

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H16F2N8
CAS Number:
Molecular Weight:
394.38
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

room temp

SMILES string

FC(F)CN(N=C1C2=CC(C=CN3)=C3N=C2)C=C1C4=NC(NCCC#N)=NC=C4

InChI

1S/C19H16F2N8/c20-16(21)11-29-10-14(15-3-7-25-19(27-15)24-5-1-4-22)17(28-29)13-8-12-2-6-23-18(12)26-9-13/h2-3,6-10,16H,1,5,11H2,(H,23,26)(H,24,25,27)

InChI key

HFYMVDKBZONUOJ-UHFFFAOYSA-N

Biochem/physiol Actions

PF-04880594 is a potent and selective RAF inhibitor that inhibits both wild-type and mutant BRAF and CRAF. PF-04880594 potently inhibits tumor growth in BRAF-mutant melanoma xenograft models.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service