Skip to Content
Merck
All Photos(1)

Key Documents

PZ0240

Sigma-Aldrich

(S)-Crizotinib

≥98% (HPLC)

Synonym(s):

3-[(1S)-1-(2,6-Dichloro-3-fluorophenyl)ethoxy]-5-[1-(4-piperidinyl)-1H-pyrazol-4-yl]-2-pyridinamine

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€103.00

€103.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€103.00

About This Item

Empirical Formula (Hill Notation):
C21H22Cl2FN5O
CAS Number:
Molecular Weight:
450.34
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

€103.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 5 mg/mL, clear (warmed)

storage temp.

room temp

SMILES string

ClC1=C([C@H](C)OC2=C(N)N=CC(C3=CN(C4CCNCC4)N=C3)=C2)C(Cl)=C(F)C=C1

InChI

1S/C21H22Cl2FN5O/c1-12(19-16(22)2-3-17(24)20(19)23)30-18-8-13(9-27-21(18)25)14-10-28-29(11-14)15-4-6-26-7-5-15/h2-3,8-12,15,26H,4-7H2,1H3,(H2,25,27)/t12-/m0/s1

InChI key

KTEIFNKAUNYNJU-LBPRGKRZSA-N

Biochem/physiol Actions

(S)-Crizotinib has a distinctly different profile from its (R)-enantiomer, the clinically approved anticancer agent Xalkori. Instead of targeting ALK, MET and ROS1, (S)-Crizotinib is a highly selective inhibitor of human 7,8-Dihydro-8-oxoguaninetriphosphatase MTH1 (NUDT1) with an IC50 value of 330 nM, 16-fold higher affinity towards MTH1 compared to the clinically used (R)-isomer. MTH1 hydrolyzes oxidized purine nucleoside triphosphates that might otherwise be incorporated into DNA/RNA and contribute to DNA damage. MTH1 removal of oxidized nucleotides that result from increased levels of reactive oxygen species (ROS) in fast-proliferating cancer cells helps protect cancer cells from proliferative stress and prevent cancer cell death. It is considered a new target for cancer therapy. In mouse xenograft studies using SW480 cells, (S)-crizotinib but not the (R)-enantiomer, was able to impair overall tumour progression as well as specifically reduce tumour volume by more than 50%.
(S)-Crizotinib is a highly selective inhibitor of human 7,8-Dihydro-8-oxoguaninetriphosphatase MTH1 (NUDT1).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Huaping Chen et al.
International journal of molecular sciences, 21(22) (2020-11-27)
MTH1 (MutT homolog 1) or NUDT1 (Nudix Hydrolase 1), also known as oxidized purine nucleoside triphosphatase, has potential as a biomarker for monitoring cancer progression and quantifying target engagement for relevant therapies. In this study, we validate one MTH1 inhibitor
Xuanxuan Dai et al.
Journal of experimental & clinical cancer research : CR, 36(1), 120-120 (2017-09-09)
Non-small cell lung cancer (NSCLC) accounts for approximately 80-85% of all lung cancers and is usually diagnosed at an advanced stage with poor prognosis. Targeted therapy has produced unprecedented outcomes in patients with NSCLC as a number of oncogenic drivers
Peng Liu et al.
Nature communications, 10(1), 1486-1486 (2019-04-04)
Immunogenic cell death (ICD) converts dying cancer cells into a therapeutic vaccine and stimulates antitumor immune responses. Here we unravel the results of an unbiased screen identifying high-dose (10 µM) crizotinib as an ICD-inducing tyrosine kinase inhibitor that has exceptional antineoplastic
Melike Çağlayan et al.
Nature communications, 8, 14045-14045 (2017-01-10)
Oxidative stress in cells can lead to accumulation of reactive oxygen species and oxidation of DNA precursors. Oxidized purine nucleotides can be inserted into DNA during replication and repair. The main pathway for correcting oxidized bases in DNA is base
Stefan Nagel et al.
Oncotarget, 11(34), 3208-3226 (2020-09-15)
NKL homeobox genes encode developmental transcription factors and display an NKL-code according to their physiological expression pattern in hematopoiesis. Here, we analyzed public transcriptome data from primary innate lymphoid cells (ILCs) for NKL homeobox gene activities and found that ILC3

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service