Skip to Content
Merck
All Photos(3)

Key Documents

PRS4053

Sigma-Aldrich

Anti-VISA (ab1) antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-CARD adapter inducing interferon-beta, Anti-MAVS, Anti-Mmitochondrial antiviral signaling protein, Anti-Virus-induced signaling adapter

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€362.00

€362.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μG
€362.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€362.00


Please contact Customer Service for Availability

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

mouse, human, rat

technique(s)

immunofluorescence: suitable
immunohistochemistry: suitable
indirect ELISA: suitable
western blot: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MAVS(57506)

Related Categories

General description

VISA (virus-induced signalling adaptor) is also designated as MAVS (mitochondrial antiviral signalling protein). It has 540 amino acids and three main domains such as N-terminal caspase recruitment and activation domain (CARD), an internal proline-rich region and a C-terminal transmembrane (TM) domain. It is mapped to chromosome 20p13. It is intermediately expressed in all tissues and brain regions except in fetal brain.

Immunogen

a 13 amino acid peptide from near the amino-terminus of human VISA.

Biochem/physiol Actions

VISA (virus-induced signalling adaptor) serves as an adaptor molecule for retinoic acid-inducible gene-I (RIG-I)-like receptors (RLRs). It activates intracellular signaling cascades, that leads to nuclear translocation of the transcription factors- IRF3 (interferon regulatory factor 3), IRF7 (interferon regulatory factor 7), and NF-κB (Nuclear factor-κB). It also helps in the formation of type I interferons (IFN-I) and proinflammatory cytokines.

Linkage

The action of this antibody can be blocked using blocking peptide SBP4053.

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

VISA--a pass to innate immunity
Qi B, et al.
The International Journal of Biochemistry & Cell Biology, 39(2), 287-291 (2007)
Activation of the RLR/MAVS Signaling Pathway by the L Protein of Mopeia Virus
Zhang LK, et al.
Journal of Virology, 90(22), 10259-10270 (2016)
Alteration of Antiviral Signalling by Single Nucleotide Polymorphisms (SNPs) of Mitochondrial Antiviral Signalling Protein (MAVS)
Xing F, et al.
PLoS ONE, 11(3) (2016)
Ling Xu et al.
Journal of immunology (Baltimore, Md. : 1950), 205(8), 2091-2099 (2020-09-11)
Hepatitis C virus (HCV) infection is the cause of severe liver disease in many people. The restricted species tropism of HCV hinders the research and development of drugs and vaccines. The Chinese tree shrew (Tupaia belangeri chinensis) is a close

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service