Skip to Content
Merck
All Photos(1)

Key Documents

PRS3765

Sigma-Aldrich

Anti-SOCS1 (ab1) antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-JAB, Anti-JAK-binding protein, Anti-Suppressor of cytokine signaling 1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€411.00

€411.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μG
€411.00

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

€411.00


Please contact Customer Service for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

predicted mol wt 23 kDa

species reactivity

human, rat, mouse

technique(s)

indirect ELISA: suitable
western blot: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SOCS1(8651)

Immunogen

a 15 amino acid peptide from near the carboxy-terminus of human SOCS1.

Linkage

The action of this antibody can be blocked using blocking peptide SBP3765.

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhelong Li et al.
Nano letters, 19(1), 19-28 (2018-12-06)
In vitro and in vivo delivery of RNAs of interest holds promise for gene therapy. Recently, exosomes are considered as a kind of rational vehicle for RNA delivery, especially miRNA and/or siRNA, while the loading efficiency is limited. In this

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service