PLAK0512
BRCA1 and Estrogen Receptor Alpha PLA® Antibody Kit
Powered by Bethyl Laboratories, Inc.
Select a Size
€871.00
Select a Size
About This Item
€871.00
Recommended Products
grade
Powered by Bethyl Laboratories, Inc.
technique(s)
proximity ligation assay: suitable (direct)
shipped in
wet ice
storage temp.
2-8°C
Application
Linkage
Analysis Note
Legal Information
Disclaimer
Storage Class Code
12 - Non Combustible Liquids
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service