Skip to Content
Merck
All Photos(1)

Key Documents

PLA0294

Sigma-Aldrich

Goat anti-H2AX Antibody, Affinity Purified

Powered by Bethyl Laboratories, Inc.

Synonym(s):

H2A histone family, H2A.X, H2A/X, H2AX, H2AX histone, histone H2A.x, member X

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€469.00

€469.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μL
€469.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€469.00


Please contact Customer Service for Availability

biological source

goat

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

species reactivity

human, mouse

technique(s)

immunohistochemistry: 1:1,000-1:5,000
western blot: 1:1,000- 1:5,000

accession no.

P16104

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

goat ... H2AX(3014)

Immunogen

The epitope recognized by PLA0294 maps to the C-terminus of human Histone H2AX using the numbering given in entry P16104 (GeneID 3014).

Physical form

Tris-citrate/phosphate buffer, pH 7 to 8 containing 0.09% Sodium Azide

Other Notes

H2AX is a member of the histone H2A family. The four core histones involved in the formation of the nucleosome structure of compacted chromatin are H2A, H2B, H3, and H4. H2AX may function to facilitate DNA repair, and recent studies have shown that H2AX is required for the maintenance of genomic stability. Gamma H2AX is the phosphorylated form of H2AX that results in response to DNA damage.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andrew M Cobb et al.
Arteriosclerosis, thrombosis, and vascular biology, 41(4), 1339-1357 (2020-12-29)
[Figure: see text].

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service