Skip to Content
Merck
All Photos(2)

Key Documents

PLA0226

Sigma-Aldrich

Rabbit anti-CKII beta Antibody, Affinity Purified

Powered by Bethyl Laboratories, Inc.

Synonym(s):

CK II beta, CK2B, CK2N, CSK2B, Casein kinase II beta subunit, G5A, G5a, beta polypeptide, casein kinase 2, phosvitin, protein G5a

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

grade

Powered by Bethyl Laboratories, Inc.

species reactivity

human, mouse

technique(s)

immunoprecipitation (IP): 2-5 μg/mg
western blot: 1:2,000- 1:10,000

accession no.

NP_001311.3

shipped in

wet ice

storage temp.

2-8°C

Gene Information

human ... CSNK2B(1460)

Immunogen

The epitope recognized by PLA0226 maps to a region between residue 165 and 215 of human casein kinase 2, beta using the numbering given in entry NP_001311.3 (GeneID 1460).

Physical form

Tris-buffered Saline containing 0.1% BSA containing 0.09% Sodium Azide

Other Notes

CKII beta (casein kinase II subunit beta) is the beta subunit of the heterotetrameric kinase, casein kinase II (CKII) which is composed of an alpha subunit, an alpha′ subunit, and two beta subunits. CKII localizes to the endoplasmic reticulum and the Golgi apparatus and is involved in signal transduction and cell cycle progression.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service