PLA0111
Rabbit anti-Lasu1/Ureb1 Antibody, Affinity Purified
Powered by Bethyl Laboratories, Inc.
Synonym(s):
ARF-BP1, ARF-binding protein 1, BJ-HCC-24 tumor antigen, HECT, HECT domain protein LASU1, HECTH9, HSPC272, Ib772, KIAA0312, LASU1, MULE, Mcl-1 ubiquitin ligase E3, UBA and WWE domain containing 1, UBA and WWE domain-containing protein 1, URE-B1, URE-binding protein 1, UREB1, homologous to E6AP carboxyl terminus homologous protein 9, large structure of UREB1, upstream regulatory element-binding protein 1
Select a Size
Select a Size
About This Item
biological source
rabbit
Quality Level
antibody form
affinity purified immunoglobulin
antibody product type
primary antibodies
grade
Powered by Bethyl Laboratories, Inc.
species reactivity
human
technique(s)
immunohistochemistry: 1:1000-1:5,000
immunoprecipitation (IP): 2-10 μg/mg
western blot: 1:2,000-1:10,000
accession no.
NP_113584.3
shipped in
wet ice
storage temp.
2-8°C
target post-translational modification
unmodified
Gene Information
rabbit ... Lasu1/Ureb1(10075)
Immunogen
Physical form
Other Notes
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
12 - Non Combustible Liquids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service