Skip to Content
Merck
All Photos(1)

Key Documents

P9649

Sigma-Aldrich

2-Oleoyl-1-stearoyl-sn-glycero-3-phosphocholine

~97% (positional purity of fatty acids), ~99% (TLC)

Synonym(s):

1-Octadecanoyl-2-(cis-9-Octadecenoyl)-sn-glycero-3-phosphocholine, L-α-Phosphatidylcholine, β-oleoyl-γ-stearoyl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C44H86NO8P
CAS Number:
Molecular Weight:
788.13
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:

Assay

~97% (positional purity of fatty acids)
~99% (TLC)

functional group

phospholipid

shipped in

ambient

storage temp.

−20°C

SMILES string

CCCCCCCCCCCCCCCCCC(=O)OC[C@@H](COP(O)(=O)OCC[N](C)(C)C)OC(=O)CCCCCCC\C=C/CCCCCCCC

InChI

1S/C44H87NO8P/c1-6-8-10-12-14-16-18-20-22-24-26-28-30-32-34-36-43(46)50-40-42(41-52-54(48,49)51-39-38-45(3,4)5)53-44(47)37-35-33-31-29-27-25-23-21-19-17-15-13-11-9-7-2/h21,23,42H,6-20,22,24-41H2,1-5H3,(H,48,49)/b23-21-

InChI key

MEHJQKIXLXVPCO-LNVKXUELSA-N

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sumit Garg et al.
Biophysical journal, 92(4), 1263-1270 (2006-11-23)
The degree of domain registration in a liquid-ordered/liquid-disordered phase-separating lipid mixture consisting of 1-stearoyl-2-oleoyl-sn-3-phosphocholine, egg sphingomyelin, and cholesterol (molar mixing ratio of 1:1:1) was studied using three different planar lipid bilayer architectures distinguished by their bilayer-substrate distance d using epifluorescence
Feng Gao et al.
Journal of the American Chemical Society, 128(14), 4814-4822 (2006-04-06)
Vesicles prepared by DMPC (1,2-dimyristoyl-sn-glycero-3-phosphocholine) and SOPC (1-stearoyl-2-oleoyl-sn-glycero-3-phosphocholine) lipid molecules having sizes smaller than the diffraction-limited focused laser beam have been used to confine single molecules in the laser focus. The confinement of single molecules in a volume smaller than
Jianjun Pan et al.
Physical review letters, 100(19), 198103-198103 (2008-06-04)
Cholesterol is well known to modulate the physical properties of biomembranes. Using modern x-ray scattering methods, we have studied the effects of cholesterol on the bending modulus K(C), the thickness D(HH), and the orientational order parameter S(xray) of lipid bilayers.
Alexander I Greenwood et al.
Biochimica et biophysica acta, 1778(4), 1120-1130 (2008-02-12)
This study uses low-angle (LAXS) and wide-angle (WAXS) X-ray synchrotron scattering, volume measurements and thin layer chromatography to determine the structure and interactions of SOPC, SOPC/cholesterol mixtures, SOPC/peptide and SOPC/cholesterol/peptide mixtures. N-acetyl-LWYIK-amide (LWYIK) represents the naturally-occurring CRAC motif segment in
Trang T Nguyen et al.
Analytical chemistry, 83(15), 5979-5988 (2011-06-24)
Here we report the use of counter-propagating second harmonic generation (SHG) to image the interactions between the local anesthetic tetracaine and a multicomponent planar supported lipid bilayer array in a label-free manner. The lipid bilayer arrays, prepared using a 3D

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service