Skip to Content
Merck
All Photos(2)

Key Documents

P9497

Sigma-Aldrich

Anti-Potassium Channel hKv11.1 antibody produced in rabbit

affinity isolated antibody, lyophilized powder

Synonym(s):

Anti-Ether-a-go-go-related Channel, Anti-HERG

Sign Into View Organizational & Contract Pricing

Select a Size

200 μL
€841.00

€841.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
200 μL
€841.00

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

€841.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

human

technique(s)

western blot: 1:400 using the lysate of HEK 293 cells, stably expressing HERG

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... KCNH2(3757)
mouse ... Kcnh2(16511)
rat ... Kcnh2(117018)

Immunogen

GST fusion protein with sequence corresponding to residues 1106-1159 of human KV11.1. The sequence is identical in rabbit and has 51/54, 50/54, and 50/54 residues identical in dog, mouse, and rat. Homology with related proteins: rat erg2 is 22/54 residues identical and rat erg3 is 21/54 residues identical.

Physical form

Lyophilized powder from phosphate buffered saline containing 1% bovine serum albumin and 0.025% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sandra Zampieri et al.
Diagnostics (Basel, Switzerland), 11(10) (2021-10-24)
The potassium channel encoded by the ether-a-gogo-related gene 1A (erg1a) has been detected in the atrophying skeletal muscle of mice experiencing either muscle disuse or cancer cachexia and further evidenced to contribute to muscle deterioration by enhancing ubiquitin proteolysis; however
Clayton Whitmore et al.
Skeletal muscle, 10(1), 1-1 (2020-01-18)
Skeletal muscle atrophy is the net loss of muscle mass that results from an imbalance in protein synthesis and protein degradation. It occurs in response to several stimuli including disease, injury, starvation, and normal aging. Currently, there is no truly

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service