Skip to Content
Merck
All Photos(1)

Key Documents

P8880

Sigma-Aldrich

trans-Pyridine-2-azo-p-dimethylaniline

Synonym(s):

2-(p-[Dimethylamino]phenylazo)pyridine: PADA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H14N4
CAS Number:
Molecular Weight:
226.28
EC Number:
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:

mp

108-110 °C (lit.)

SMILES string

CN(C)c1ccc(cc1)N=Nc2ccccn2

Looking for similar products? Visit Product Comparison Guide

Application

Reagent for the qualitative and quantitative determination of −SH groups.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A spectrophotometric titration for the determination of sulfhydryl groups.
I M KLOTZ et al.
Archives of biochemistry and biophysics, 95, 540-546 (1961-12-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service