Skip to Content
Merck
All Photos(1)

Key Documents

P8664

Sigma-Aldrich

1,2-Diheptadecanoyl-sn-glycero-3-phosphoethanolamine

≥99% (GC)

Synonym(s):

L-α-Phosphatidylethanolamine, diheptadecanoyl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C39H78NO8P
CAS Number:
Molecular Weight:
720.01
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:

Assay

≥99% (GC)

functional group

amine

shipped in

ambient

storage temp.

−20°C

SMILES string

CCCCCCCCCCCCCCCCC(=O)OCC(COP(O)(=O)OCCN)OC(=O)CCCCCCCCCCCCCCCC

InChI

1S/C39H78NO8P/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-38(41)45-35-37(36-47-49(43,44)46-34-33-40)48-39(42)32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h37H,3-36,40H2,1-2H3,(H,43,44)

InChI key

YSFFAUPDXKTJMR-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhenwen Zhao et al.
Lipids, 51(2), 179-187 (2015-12-15)
Despite huge advances in the research of epithelial ovarian cancer (EOC), it remains the most lethal gynecological malignancy. Peritoneal tumor cell dissemination with cell survival and drug-resistance to taxane and platinum-based chemotherapy are two of the major challenges of EOC
Chao Han et al.
The Analyst, 144(21), 6304-6312 (2019-09-26)
Matrix-assisted laser desorption/ionization mass spectrometry imaging (MALDI MSI) is a powerful tool for the characterization and localization of analytes without the need for extraction, purification, separation or labeling of samples. However, in tissue sections the most abundant lipids, phosphatidylcholines (PCs)
Hye-Youn Kim et al.
Scientific reports, 9(1), 3140-3140 (2019-03-01)
Melanoma is the most aggressive form of skin cancer, with metastatic melanoma being refractory to currently available conventional therapies. In this study, we evaluated the inhibitory effect of coronatine (COR) on the proliferation of metastatic melanoma cells. COR inhibited the
Santosh Lamichhane et al.
Scientific data, 5, 180250-180250 (2018-11-14)
Early prediction and prevention of type 1 diabetes (T1D) are currently unmet medical needs. Previous metabolomics studies suggest that children who develop T1D are characterised by a distinct metabolic profile already detectable during infancy, prior to the onset of islet
Aidan McGlinchey et al.
Environment international, 143, 105935-105935 (2020-07-08)
In the last decade, increasing incidence of type 1 diabetes (T1D) stabilized in Finland, a phenomenon that coincides with tighter regulation of perfluoroalkyl substances (PFAS). Here, we quantified PFAS to examine their effects, during pregnancy, on lipid and immune-related markers

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service