Skip to Content
Merck
All Photos(1)

Key Documents

P8018

Sigma-Aldrich

Potassium hexachloroplatinate(IV)

powder

Synonym(s):

Dipotassium hexachloroplatinate, Dipotassium platinum hexachloride, Platinic potassium chloride, Platinum potassium chloride, Potassium chloroplatinate, Potassium platinum(IV) chloride, Potassium platinum(IV) hexachloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
K2PtCl6
CAS Number:
Molecular Weight:
485.99
EC Number:
MDL number:
UNSPSC Code:
12352103

form

powder

color

yellow

mp

250 °C (dec.) (lit.)

SMILES string

[K+].[K+].Cl[Pt--](Cl)(Cl)(Cl)(Cl)Cl

InChI

1S/6ClH.2K.Pt/h6*1H;;;/q;;;;;;2*+1;+4/p-6

InChI key

DPAIVKJGTXERIM-UHFFFAOYSA-H

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Met. Corr. 1 - Resp. Sens. 1 - Skin Sens. 1 - STOT RE 1

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service