Skip to Content
Merck
All Photos(1)

Key Documents

P7679

Sigma-Aldrich

Monoclonal Anti-Platelet-Derived Growth Factor Receptor β antibody produced in mouse

clone PDGFR-B2, ascites fluid

Synonym(s):

Anti-PDGF Rβ

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
51111800

biological source

mouse

conjugate

unconjugated

antibody form

ascites fluid

clone

PDGFR-B2, monoclonal

contains

15 mM sodium azide

species reactivity

human, pig

technique(s)

immunohistochemistry (frozen sections): suitable
immunoprecipitation (IP): suitable
microarray: suitable
western blot: 1:100 using denatured, non-reduced porcine uterus extract

isotype

IgG2b

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... PDGFRB(5159)

Specificity

The antibody recognizes human and pig PDGF Rβ, but not mouse or rat. It can be used for immunoperoxidase and immunofluorescence staining of unfixed or acetone-fixed frozen tissue. The antibody induces clustering and down-regulation of PDGF Rβ. The antibody does not cross react with PDGF Rα.

Immunogen

PDGF β-receptor from pig uterus.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C Reuterdahl et al.
Laboratory investigation; a journal of technical methods and pathology, 64(3), 321-329 (1991-03-01)
Platelet-derived growth factor (PDGF) beta-receptor expression in normal and rheumatoid synovia was investigated by double immunofluorescence staining of frozen sections and by in situ hybridization. In the inflamed synovia, PDGF beta-receptor mRNA was present in vascular cells, as well as
A Sorkin et al.
The Journal of cell biology, 112(3), 469-478 (1991-02-01)
The complementary DNAs for wildtype and tyrosine kinase-inactivated (K634A) forms of the PDGF beta-receptor were expressed in porcine aortic endothelial cells. We examined the internalization and degradation of ligands and receptors after exposure of receptor expressing cells to PDGF-BB, which
P Höddelius et al.
Bioscience reports, 20(2), 119-127 (2000-08-31)
When the receptors for platelet-derived growth factor (PDGF) are activated they aggregate, become tyrosine-phosphorylated and elicit a cascade of down-stream signals, including mobilization of Ca2+ from intra- and extracellular stores. Receptor mobility in the plane of the membrane is a
Mikhail Sergeev et al.
Biophysical journal, 91(10), 3884-3896 (2006-08-29)
We present higher-order moment analysis of fluorescence intensity fluctuations from individual laser scanning microscopy images applied to study monomer-oligomer distributions. We demonstrate that the number densities and brightness ratios of a mixed population of monomers and oligomers can be determined
Jonathan W D Comeau et al.
Biophysical journal, 91(12), 4611-4622 (2006-10-03)
Biomolecular interactions are fundamental to the vast majority of cellular processes, and identification of the major interacting components is usually the first step toward an understanding of the mechanisms that govern various cell functions. Thus, statistical image analyses that can

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service