P7348
T7 Phage Promoter Primer Set
Synonym(s):
5′-TAA TAC GAC TCA CTA TAG GG-3′
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
storage temp.
2-8°C
Application
T7 Phage Promoter Primer Set is a single-stranded oligonucleotide with 5′-hydroxyl and 3′-hydroxyl ends and a selection of four fluorescent lables for use in polymerase chain reaction protocols (PCR). Functionally tested for use in fluorescence-detection automated sequencing.
Packaging
Packaged in amber tubes to protect from light.
Components
Set of four vials of primer, each individually labeled at the 5′-end with:
FAM: (5[6]-carboxyfluorescein)
JOE: (6-carboxy-4′,5′-dichloro-2′,7′-dimethoxyfluorescein)
ROX: (5[6]-carboxy-X-rhodamine), and
TAMRA: (5[6]-carboxytetramethylrhodamine.
Supplied as solutions, 50 μl each, in tris-EDTA buffer, pH 7.5, at 1 picomole/μl
FAM: (5[6]-carboxyfluorescein)
JOE: (6-carboxy-4′,5′-dichloro-2′,7′-dimethoxyfluorescein)
ROX: (5[6]-carboxy-X-rhodamine), and
TAMRA: (5[6]-carboxytetramethylrhodamine.
Supplied as solutions, 50 μl each, in tris-EDTA buffer, pH 7.5, at 1 picomole/μl
Analysis Note
Tested for purity with HPLC, PAGE, and OD.
Storage Class Code
10 - Combustible liquids
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
PloS one, 6(7), e22224-e22224 (2011-07-27)
Eukaryotic DNA polymerase δ (pol δ) plays a crucial role in chromosomal DNA replication and various DNA repair processes. It is thought to consist of p125, p66 (p68), p50 and p12 subunits. However, rigorous isolation of mammalian pol δ from
High-throughput sequencing of PCR products tagged with universal primers using 454 life sciences systems.
Current Protocols in Molecular Biology, Unit 7-Unit 7 (2011)
Journal of virological methods, 178(1-2), 253-257 (2011-10-04)
Human norovirus (HuNoV) and hepatitis A (HAV) are recognized as leading causes of non-bacterial foodborne associated illnesses in the United States. DNA sequencing is generally considered the standard for accurate viral genotyping in support of epidemiological investigations. Due to the
Genetics and molecular research : GMR, 9(4), 2016-2024 (2010-10-20)
Seven fungal isolates were identified as pan-global Hypocrea/Trichoderma species, from section Trichoderma, on the basis of their morphology. These species were H. lixii/T. harzianum and H. orientalis/T. longibrachiatum. PCR-based markers with primer M13 (core sequence of phage M13) and internal-transcribed
International journal of food microbiology, 144(1), 187-192 (2010-10-12)
The present research studied Saccharomyces cerevisiae yeasts isolated from Nero d'Avola grapes, collected in different areas of the Sicily region. RAPD-PCR analysis with M13 primer was used for preliminary discrimination among 341 S. cerevisiae isolates. Inoculated fermentations with S. cerevisiae
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service