Key Documents
P6983
Hamilton® pipette tips
Filter Tips, maximum volume 1 mL
About This Item
UNSPSC Code:
41121601
NACRES:
NB.01
Pricing and availability is not currently available.
material
colorless
description
for Softgrip pipettes
sterility
sterile
feature
barrier
packaging
pack of 576 ea (6 racks of 96 tips)
manufacturer/tradename
Hamilton 11003-09
maximum volume
1 mL
Other Notes
Browse all pipette tips
Legal Information
Hamilton is a registered trademark of Hamilton Co.
Choose from one of the most recent versions:
Certificates of Analysis (COA)