Skip to Content
Merck

Skip To

P6972

Monoclonal Anti-Interferon-γ−R-Phycoerythrin antibody produced in mouse

clone 25723, purified immunoglobulin, saline solution

Synonym(s):

Monoclonal Anti-Interferon-γ

Sign In to View Organizational & Contract Pricing.

About This Item

NACRES:
NA.41
UNSPSC Code:
51111800
Conjugate:
phycoerythrin (R-PE) conjugate
Clone:
25723, monoclonal
Application:
FACS
Citations:
1


biological source

mouse

Quality Segment

conjugate

phycoerythrin (R-PE) conjugate

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

25723, monoclonal

form

saline solution

species reactivity

human

technique(s)

flow cytometry: suitable

isotype

IgG2b

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

Gene Information

human ... IFNG(3458)

General description

Human Interferon-γ (IFN-γ) is a Type I cytokine produced by the lymphocytes activated in response to specific antigens or mitogens. It has a pivotal role in activating primed macrophages for nonspecific tumoricidal activity as well as can stimulate monocytes for enhanced cytotoxicity against tumor cells. It also has several biological functions like inhibition of cell or tumor growth, promotion of B cell differentiation into plasma cells and antiviral activity. Further, it can enhance the cytotoxicity of natural killer cells and T-cells. Monoclonal anti-Interferon-γ−R-Phycoerythrin antibody reacts specifically with human IFN-γ.

Immunogen

recombinant human IFN-γ

Application

Monoclonal anti-Interferon-γ−R-Phycoerythrin antibody can be used in flow cytometry for detecting intracytoplasmic cytokines and enumerating cytokine-producing cells in a mixed population.

Biochem/physiol Actions

The conjugates will identify and quantitate cells containing intracellular forms of human IFN-γ by flow cytometry.

Physical form

Solution in saline containing up to 0.5% bovine serum albumin and 0.1% sodium azide as a preservative.

Preparation Note

Purified antibody is conjugated to R-phycoerythrin (PE).

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters