Skip to Content
Merck
All Photos(1)

Key Documents

P6889

Sigma-Aldrich

Propionaldehyde

Synonym(s):

Propanal

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CHO
CAS Number:
Molecular Weight:
58.08
Beilstein:
506010
EC Number:
MDL number:
UNSPSC Code:
12352103

vapor density

2 (vs air)

vapor pressure

18.77 psi ( 55 °C)
4.89 psi ( 20 °C)

autoignition temp.

404 °F

expl. lim.

17 %, 26 °F
2.6 %, 31 °F

refractive index

n20/D 1.362 (lit.)

bp

46-50 °C (lit.)

mp

−81 °C (lit.)

density

0.805 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

[H]C(=O)CC

InChI

1S/C3H6O/c1-2-3-4/h3H,2H2,1H3

InChI key

NBBJYMSMWIIQGU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cynthia A Morgan et al.
Chemico-biological interactions, 234, 29-37 (2014-12-03)
The human aldehyde dehydrogenase (ALDH) superfamily consists of at least 19 enzymes that metabolize endogenous and exogenous aldehydes. Currently, there are no commercially available inhibitors that target ALDH1A1 but have little to no effect on the structurally and functionally similar
Michela Salamone et al.
The Journal of organic chemistry, 76(11), 4645-4651 (2011-04-30)
A kinetic study of the hydrogen atom abstraction reactions from propanal (PA) and 2,2-dimethylpropanal (DMPA) by the cumyloxyl radical (CumO•) has been carried out in different solvents (benzene, PhCl, MeCN, t-BuOH, MeOH, and TFE). The corresponding reactions of the benzyloxyl
Chenguang Fan et al.
Proceedings of the National Academy of Sciences of the United States of America, 109(37), 14995-15000 (2012-08-29)
Bacterial microcompartments (MCPs) are a widespread family of proteinaceous organelles that consist of metabolic enzymes encapsulated within a protein shell. For MCPs to function specific enzymes must be encapsulated. We recently reported that a short N-terminal targeting sequence of propionaldehyde
Lian Hua Luo et al.
Applied microbiology and biotechnology, 89(3), 697-703 (2010-10-05)
Although the de novo biosynthetic mechanism of 3-hydroxypropionic acid (3-HP) in glycerol-fermenting microorganisms is still unclear, the propanediol utilization protein (PduP) of Lactobacillus species has been suggested to be a key enzyme in this regard. To verify this hypothesis, a
Lian Hua Luo et al.
Bioresource technology, 103(1), 1-6 (2011-10-25)
The pduP gene encodes a propionaldehyde dehydrogenase (PduP) was investigated for the role in 3-hydroxypropionic acid (3-HP) glycerol metabolism in Klebsiella pneumoniae. The enzyme assay showed that cell extracts from a pduP mutant strain lacked measurable dehydrogenase activity. Additionally, the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service