Skip to Content
Merck
All Photos(1)

Key Documents

P4360

Sigma-Aldrich

Monoclonal Anti-p107 antibody produced in mouse

clone KAB6, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-CP107, Anti-PRB1, Anti-p107

Sign Into View Organizational & Contract Pricing

Select a Size

0.1 MG
€508.00

€508.00


Please contact Customer Service for Availability


Select a Size

Change View
0.1 MG
€508.00

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

€508.00


Please contact Customer Service for Availability

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

KAB6, monoclonal

form

buffered aqueous solution

mol wt

antigen 107 kDa

species reactivity

mouse, human

technique(s)

western blot: 1:1,000 using human C33A cervical carcinoma cells

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RBL1(5933)
mouse ... Rbl1(19650)

Immunogen

GST fusion protein of p107 N-terminus.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing and 0.08% sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nan Wu et al.
The Journal of clinical investigation, 127(3), 888-898 (2017-02-07)
The most frequent focal alterations in human retinoblastoma are mutations in the tumor-suppressor gene retinoblastoma (RB) and amplification of the oncogene MYCN. Whether MYCN overexpression drives retinoblastoma has not been assessed in model systems. Here, we have shown that Rb

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service